Italy
Order Entry
Filter tips, Biosphere®
 :  SARSTEDT
Filter tips, Biosphere®
 :  SARSTEDT
  •  
    Tip style
    Volume
    Sterile
    Tip colour
    For use with
    Packed
  • SARS70.3010.255
    70.3010.255
    Type E
    10 µl
    +
    Transparent
    Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, eLine Electronic
    96/box, 480/inner box
    1 * 1.920 pezzi
    1 * 1.920 pezzi - 279,00 €
    279,00 €
    1 * 1.920 pezzi
       279,00 €