Italy
Order Entry
VWR® AVS TITRINORM, Thallium Standard Solution, 1000 mg/L Tl in dil. Nitric Acid, Standard for AAS
VWR® AVS TITRINORM, Thallium Standard Solution, 1000 mg/L Tl in dil. Nitric Acid, Standard for AAS
Supplier:  VWR Chemicals
Certificates
undefined
VWR® AVS TITRINORM, Thallium Standard Solution, 1000 mg/L Tl in dil. Nitric Acid, Standard for AAS
Supplier:  VWR Chemicals
Certificates
Related Information
Product Information
  • Catalog #
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
    Availability
    Price
  • Thallium
    Tl
    1000 mg/L
    Nitric acid dil.
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 34,60 €
    34,60 €
    1 * 100 mL
       34,60 €
    Local Article Number: VWRC86718.180
  • Thallium
    Tl
    1000 mg/L
    Nitric acid dil.
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 57,30 €
    57,30 €
    1 * 500 mL
       57,30 €
    Local Article Number: VWRC86718.260