Stampa…
Hai cercato: Piastre di estrazione in fase solida
53  risultati sono stati trovati
SearchResultCount:"53"
Sort Results
Visualizzazione a lista Nuova Visualizzazione
La tua valutazione
Fornitore: Avantor
Descrizione: Solid Phase Extraction (SPE) is commonly used as a highly selective sample clean-up or sample enrichment method prior to subsequent analyses. J.T.Bake...
Codice catalogo: (TCAT613-0151RCH)
Fornitore: Tecan
Codice articolo fornitore: 613-0151RCH
Codice articolo locale: TCAT613-0151RCH
Descrizione: A 10 µm silica micro particulate with polar C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform...
UOM: 1 * 1 pezzi
New Product
Codice catalogo: (TCAT694-0201RCH)
Fornitore: Tecan
Codice articolo fornitore: 694-0201RCH
Codice articolo locale: TCAT694-0201RCH
Descrizione: A 18 µm polymer micro particulate with weak anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fas...
UOM: 1 * 1 pezzi
New Product
Fornitore: Tecan
Descrizione: A 25 µm polymer micro particulate with weak cationic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fa...
New Product
Fornitore: Tecan
Descrizione: A hydrophilic/lipophilic balanced strong cationic chemistry offering excellent selectivity to basic and neutral compounds. A direct alternative to wat...
New Product
Codice catalogo: (TCAT642-0501RCH)
Fornitore: Tecan
Codice articolo fornitore: 642-0501RCH
Codice articolo locale: TCAT642-0501RCH
Descrizione: A 10 µm micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and ...
UOM: 1 * 1 pezzi
New Product
Fornitore: Tecan
Descrizione: Proprietary blend of ion-exchange and water-wettable polymer targeted towards oral fluid collection devices.
New Product
Fornitore: Tecan
Descrizione: A hydrophilic/Lipophilic balanced chemistry offering selectivity to a wide range of analytes. It provides enhanced selectivity for polar species along...
New Product
Fornitore: Tecan
Descrizione: A 30 µm polymer micro particulate with strong cationic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a ...
New Product
Fornitore: Tecan
Descrizione: A hydrophilic/lipophilic balanced chemistry offering selectivity to a wide range of analytes. It provides enhanced selectivity for polar species, also...
New Product
Codice catalogo: (TCAT687-0101RCH)
Fornitore: Tecan
Codice articolo fornitore: 687-0101RCH
Codice articolo locale: TCAT687-0101RCH
Descrizione: A 25 µm polymer micro particulate with strong cationic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a ...
UOM: 1 * 1 pezzi
New Product
Codice catalogo: (TCAT417-0101RCH)
Fornitore: Tecan
Codice articolo fornitore: 417-0101RCH
Codice articolo locale: TCAT417-0101RCH
Descrizione: Special water wettable polymer with 300A pore size ideal for protein digest clean up and other protein related workflows.
UOM: 1 * 1 pezzi
New Product
Codice catalogo: (TCAT667-0101RCH)
Fornitore: Tecan
Codice articolo fornitore: 667-0101RCH
Codice articolo locale: TCAT667-0101RCH
Descrizione: A 15 µm micro particulate with strong cation exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and u...
UOM: 1 * 1 pezzi
New Product
Codice catalogo: (TCAT675-0051NBE)
Fornitore: Tecan
Codice articolo fornitore: 675-0051NBE
Codice articolo locale: TCAT675-0051NBE
Descrizione: A 15 µm polymer micro particulate with weak cationic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fa...
UOM: 1 * 1 pezzi
New Product
Fornitore: Tecan
Descrizione: NBE CFP 96-well plates has two types of sorbent polypropylene or PTFE.
New Product
Codice catalogo: (TCAT675-0201RCH)
Fornitore: Tecan
Codice articolo fornitore: 675-0201RCH
Codice articolo locale: TCAT675-0201RCH
Descrizione: Polymeric, weak cation exchange chemistry for strong bases.
UOM: 1 * 1 pezzi
New Product
Prezzo a richiesta
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Questo prodotto è stato bloccato dalla vostra organizzazione. Si prega di contattare il vostro responsabile acquisti per ulteriori informazioni.
Il prodotto originale non è più disponibile. L'articolo suggerito è disponibile
Il prodotto marcato con questo simbolo non è più disponibile o vendibile fino ad esaurimento scorte. Alternative possono essere disponibili utilizzando il numero di catalogo VWR. Per maggiori informazioni può contattare il nostro customer service al 023320311 o inviando una email all'indirizzo ebusiness.it@vwr.com
1 - 16 of 53
no targeter for Bottom