Italy
Order Entry
PCR tube racks, PP, Axygen®
 :  Corning
undefined
PCR tube racks, PP, Axygen®
 :  Corning
  • Catalog No:
  • 732-1389
  • 732-1388
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  •  
    Colour
    No. of Holes
    For tubes
    Pk
  • 732-1389
    R-96-PCR-FY
    Hi-vis yellow
    96
    0,2/0,5 ml
    5
    1 * 5 pezzi
    1 * 5 pezzi - 54,40 €
    54,40 €
    1 * 5 pezzi
       54,40 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • 732-1388
    R-96-PCR-FSP
    Assorted
    96
    0,2/0,5 ml
    25
    1 * 25 pezzi
    1 * 25 pezzi - 273,00 €
    273,00 €
    1 * 25 pezzi
       273,00 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact