Italy
Order Entry
1,4-Diisopropylbenzene ≥98.0% (by GC)
 :  TCI
undefined
1,4-Diisopropylbenzene ≥98.0% (by GC)
 :  TCI
  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  •  
    Pack type
    Pk
  • TCIAD0928-25ML
    D0928-25ML
    Glass bottle
    25 mL
    1 * 25 mL
    1 * 25 mL - 39,00 €
    39,00 €
    1 * 25 mL
       39,00 €
     : D0928-25ML
     : TCIAD0928-25ML
  • TCIAD0928-100ML
    D0928-100ML
    100 mL
    1 * 100 mL
    1 * 100 mL - 153,00 €
    153,00 €
    1 * 100 mL
       153,00 €
     : D0928-100ML
     : TCIAD0928-100ML