Stampa…
Standard transfer tank with cooling chamber, TE62
Fornitore: Hoefer
732-2192EA 1369 EUR
HOEFTE62
Standard transfer tank with cooling chamber, TE62
The TE62 standard transfer tank with cooling chamber quickly and evenly transfers proteins and nucleic acids from polyacrylamide or agarose gels onto nylon, nitrocellulose, or PVDF membranes. The TE62 is designed to accept up to four cassettes simultaneously with cooling. Each cassette holds a 15×21 cm gel, or as many as four 7×10 cm mini gels.
  • Uniform and strong electric field supports efficient and even transfers
  • Colour coded, easy to assemble cassettes ensure proper orientation during transfer
  • Superior tank design allows the cassettes to apply equal pressure across the stack preventing gel distortion
  • Built-in ceramic heat exchanger provides excellent buffer temperature control when used with an external cooling water bath
Informazioni per l'ordine: Each system includes lower chamber with heat exchanger, safety lid with high voltage leads, 2× electrode panels, 4× cassettes, 4× foam sponges (6 mm thick), 8× foam sponges (3 mm thick), cassette hook, and 25× blotting paper sheets (14,5×21,5 cm).