Tetrabutylammonium dihydrogen phosphate
Supplier: PanReac AppliChem
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Tetra-n-butylammonium phosphate monobasic
, Tetra-n-butylammonium phosphate
, Tetrabutylammonium phosphate monobasic
, Tetra-n-butylammonium dihydrogen phosphate
, IPC-TBA-P
A1947.0025EA
332
GBP
A1947.0025
Tetrabutylammonium dihydrogen phosphate
Tetrabutylammonium dihydrogen phosphate
Formula:
C₁₆H₃₈NO₄P MW: 339.46 g/mol |
MDL Number:
MFCD00064526 CAS Number: 5574-97-0 EINECS: 226-947-1 |