Stampa…
Inoculating loops, with sphere, Sterilin®
Fornitore: Thermo Fisher Scientific
Sterilin®
391-0474EA 270 EUR
391-0474
Inoculating loops, with sphere, Sterilin®
Spatole e aghi Anse e aghi per inoculo
PS, sterile inoculation loop for fixed sample volumes at one end and sphere for streaking at the other.
  • Used in microbiological applications for dilution streaking and obtaining isolated colonies
  • By turning the square handle 90° after each streaking the sphere provides up to four sterile streaking surfaces
  • Eliminates the need for flaming
  • Sterilised by gamma irradiation