Stampa…
Alcohol meters
Fornitore: Alla France
ALLA0900FC035/20QPEA 246 EUR
ALLA0900FC035/20QP ALLA0900FC095/20QP ALLA0900FC050/20QP ALLA0900FC011/20QP ALLA0900FC000/20QP ALLA0900FC060/20QP ALLA0900FC045/20QP ALLA0900FC015/20QP ALLA0900FC080/20QP ALLA0900FC020/20QP ALLA0900FC040/20QP ALLA0900FC009/20QP ALLA0900FC070/20QP ALLA0900FC030/20QP ALLA0900FC006/20QP
Alcohol meters
Hydrometers for the determination of sugar in alcohol.
  • ° Twaddle scale
  • Available in two lengths
  • Reading below meniscus
Informazioni per l'ordine: Certificate is not included.