Stampa…
Safety Storage Cabinets for Non-Flammable Liquids and Water Polluting Substances, Düperthal
Fornitore: Düperthal
DUEH20-101046-003EA 457.1 EUR
DUEH20-101046-003
Safety Storage Cabinets for Non-Flammable Liquids and Water Polluting Substances, Düperthal
These safety storage cabinets are used for safe storage of non-flammable liquids and water polluting substances in small containers.
  • Torsion resistant wing doors with box stiffeners
  • Ventilation through air vents in the rear panel of the cabinet
  • Closed using turning handle with security lock on 3 sides
  • Magnet pocket for DIN A5 documents
  • Anti-tilt of the collection trays by locking device
  • Four adjustable bases for alignment of floor unevenness
Height-adjustable collection trays with galvanised optional material thickness of 1 and 3 mm.
Certificazioni: Conforms with Art 19, German Equipment and Product Safety Law (GS Mark).