Stampa…
Calibration certificates
Fornitore: EBRO
EBRO1020-3599EA 149 EUR InStock
EBRO1020-3599 EBRO1020-3799
Calibration certificates
Services
Modern quality assurance systems like ISO 9000, QS 9000, GxP and FDA require testing and measuring equipment checks, which also include a regular calibration of the devices.
  • Calibration is done by accredited laboratories
  • Identification and documentation of the measurement uncertainty
  • Manufacturer-independent calibration, device from other manufacturers can be calibrated
ISO calibration is an economical, fast and precise option for the fulfilment of these requirements. The results are documented in detail, including traceability information of the reference devices is called ISO certificate.
DAkkS calibration is needed for working standard measuring equipment, measuring equipment used by certified experts and for certain measurement procedures in medicine and pharmaceuticals.