Stampa…
Lanyard, Miller® Amorstop -Y- Lanyard
Fornitore: Miller by Honeywell
DALL1003232EA 233 EUR
DALL1003232
Lanyard, Miller® Amorstop -Y- Lanyard
This lanyard offers higher comfort in case of a fall. Ideal for ascending and descending a pole, tower or ladder: The user is always secured by two-way anchoring.
  • Made from very tough and robust 23 mm polyester fabric for longer durability
  • Small integrated shock absorber element ensures lower weight and easier handling
  • Lanyard made from webbing for 100% safety
Certificazioni: EN 362