Stampa…
ESD shirt, long sleeves, diamond knit, XF83
Fornitore: Fristads Kansas Group
Fristads®
113-05389EA 0 EUR
113-05389 113-05388 113-05398 113-05387 113-05397 113-05396 113-05395 113-05394 113-05393 113-05392 113-05391 113-05390
ESD shirt, long sleeves, diamond knit, XF83
Abbigliamento da lavoro
ESD shirt with long sleeves, manufactured from a knitted blend (83% cotton, 14% polyester and 3% conductive fibre), which makes it comfortable to wear. The special knitting technique produces good conductivity although the content of conductive fibre is only 3%.
  • Crew neck
  • With rib-knitted ESD bands
  • ESD symbol on sleeve
Weight: 160 g/m²
Certificazioni: The garments are tested and certified under IEC 61340.