Stampa…
Urine specimen collection tubes, with screw caps
Fornitore: Simport
SIMPT553-10AAEA 262 EUR
SIMPT553-10AA SIMPT553-10A
Urine specimen collection tubes, with screw caps
Graduated tubes with skirted free-standing base, contain a 75 mg tablet of boric acid in order to preserve urine specimens for up to 72 hours without refrigeration.
  • Amber colour tubes are available for light sensitive specimens
  • Conical bottom to facilitate sample removal
  • A white leakproof screw cap is supplied on each tube along with a fill line label