Stampa…
Hai cercato: Saccheti per guanti
20  risultati sono stati trovati
SearchResultCount:"20"
Sort Results
Visualizzazione a lista Nuova Visualizzazione
La tua valutazione
Fornitore: LABCONCO
Descrizione: This filtered glove box with fibreglass liner provides inlet and outlet HEPA filtration and a leak-tight physical barrier to protect the operator from...
Fornitore: LABCONCO
Descrizione: These glove boxes provides a leak-tight environment to perform work with contamination-sensitive materials under vacuum or in an inert environment.
Fornitore: Thermo Scientific
Descrizione: These polyethylene glove bags are used for handling air and moisture sensitive chemicals in laboratories. Also suitable for field sampling.
Fornitore: Thermo Scientific
Descrizione: These polyethylene glove bags are ideal for laboratory use and field sampling.
Codice catalogo: (BOHLV1984-08)
Fornitore: Bohlender
Codice articolo fornitore: V1984-08
Codice articolo locale: BOHLV1984-08
Descrizione: This PMMA glove box allows safe working with sensitive compounds in a controlled environment.
UOM: 1 * 1 pezzi
Fornitore: LABCONCO
Descrizione: These glove boxes with stainless steel liner provide inlet and outlet ULPA filtration and a leak-tight physical barrier to protect the operator from e...
Fornitore: LABCONCO
Descrizione: This combination glove box with fibreglass lining provides inlet and outlet HEPA or ULPA filtration and a leak-tight physical barrier to protect the o...
Fornitore: LABCONCO
Descrizione: These glove boxes with stainless steel liner provide inlet and outlet HEPA or ULPA filtration and a leak-tight physical barrier to protect the operato...
Codice catalogo: (BOHLV1755-08)
Fornitore: Bohlender
Codice articolo fornitore: V1755-08
Codice articolo locale: BOHLV1755-08
Descrizione: Designed for the handling of humidity-sensitive products in a dry atmosphere.
UOM: 1 * 1 pezzi
New Product
Codice catalogo: (BOHLV1982-08)
Fornitore: Bohlender
Codice articolo fornitore: V1982-08
Codice articolo locale: BOHLV1982-08
Descrizione: This PMMA glove box with transfer chamber allows safe working with sensitive compounds in a controlled environment.
UOM: 1 * 1 pezzi
Fornitore: LABCONCO
Descrizione: This glove box is designed to create controlled oxygen and moisture atmospheres for working with products that are easily oxidized. The glove box feat...
Codice catalogo: (BOHLV1992-08)
Fornitore: Bohlender
Codice articolo fornitore: V1992-08
Codice articolo locale: BOHLV1992-08
Descrizione: This anti-static glove box made of PC-ESD allows safe working with sensitive compounds in a controlled environment.
UOM: 1 * 1 pezzi
Codice catalogo: (135-1095)
Fornitore: LABCONCO
Codice articolo fornitore: 5220120
Codice articolo locale: LBCP5220120
Descrizione: This glove box provides a leak-tight environment for work with contamination-sensitive materials.
UOM: 1 * 1 pezzi
Codice catalogo: (BOHLV1988-08)
Fornitore: Bohlender
Codice articolo fornitore: V1988-08
Codice articolo locale: BOHLV1988-08
Descrizione: This normal pressure glove box features a robust yet lightweight aluminium frame with panels made of acrylic glass which ensures superb all-round visi...
UOM: 1 * 1 pezzi
Codice catalogo: (BELAH500260000)
Fornitore: Bel-Art Products, a Part of SP
Codice articolo fornitore: H500260000
Codice articolo locale: BELAH500260000
Descrizione: This small and portable glove box is designed for remote and field use. It is manufactured from transparent acrylic plastic (6,35 mm thick).
UOM: 1 * 1 pezzi
Codice catalogo: (135-1097)
Fornitore: LABCONCO
Codice articolo fornitore: 5220320
Codice articolo locale: LBCP5220320
Descrizione: XPert weigh boxes have inlet and outlet HEPA filters and provide a leak-tight physical barrier to protect the operator from hazardous airborne particu...
UOM: 1 * 1 pezzi
Prezzo a richiesta
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Questo prodotto è stato bloccato dalla vostra organizzazione. Si prega di contattare il vostro responsabile acquisti per ulteriori informazioni.
Il prodotto originale non è più disponibile. L'articolo suggerito è disponibile
Il prodotto marcato con questo simbolo non è più disponibile o vendibile fino ad esaurimento scorte. Alternative possono essere disponibili utilizzando il numero di catalogo VWR. Per maggiori informazioni può contattare il nostro customer service al 023320311 o inviando una email all'indirizzo ebusiness.it@vwr.com
1 - 16 of 20
no targeter for Bottom