Stampa…
Hai cercato: Standard per viscometria
20  risultati sono stati trovati
SearchResultCount:"20"
Sort Results
Visualizzazione a lista Nuova Visualizzazione
La tua valutazione
Fornitore: VWR Chemicals
Descrizione: Standard utilizzabili per calibrazione, controllo, verifica, qualificazione o validazione del metodo di strumenti di misurazione della viscosità dinam...
Fornitore: VWR Chemicals
Descrizione: These ISO 17025 and ISO 17034 certified general purpose viscosity standards provide certified kinematic viscosity, dynamic viscosity and density data ...
Fornitore: VWR Chemicals
Descrizione: High quality CCS viscosity standards are ideal for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) test...
Codice catalogo: (99566.180)
Fornitore: VWR Chemicals
Codice articolo locale: VWRC99566.180
Descrizione: Pure Water viscosity standard have certified data for kinematic viscosity, dynamic viscosity and density at 5, 20, 25 and 37 °C.
UOM: 1 * 100 mL
MSDS
Fornitore: VWR Chemicals
Descrizione: These standards are certified in strict accordance with ASTM D2162 at 40 and 100 °C under ISO 17025 and ISO 17034 accreditation, and have been manufac...
Fornitore: VWR Chemicals
Descrizione: The viscosity values for certified viscosity check oils have been tested and certified at typical blend targets. These check oils are for validating t...
Fornitore: VWR Chemicals
Descrizione: Low Temperature viscosity standards have certified data for kinematic viscosity, dynamic viscosity and density for various low temperatures.
Fornitore: VWR Chemicals
Descrizione: Mineral Oil Rotational viscosity standards are specifically formulated for use with rotational viscometers and are dual-certified to ISO 17025 and ISO...
Fornitore: VWR Chemicals
Descrizione: Medical Grade viscosity standards provide certified dynamic viscosity data at 25 and 37 °C, are dual-certified to ISO 17025 and ISO 17034 under UKAS a...
Fornitore: VWR Chemicals
Descrizione: Flow Cup viscosity standards are specifically formulated for use with viscosity flow cups such as Ford, Shell and Zahn.
Fornitore: VWR Chemicals
Descrizione: Paints and coatings viscosity standards provide certified kinematic viscosity, dynamic viscosity, density and (where applicable) Krebs data across tem...
Fornitore: VWR Chemicals
Descrizione: Silicone Rotational viscosity standards are specifically formulated for use with equipment with minimal temperature control. Supplied in 600 ml 'ready...
Fornitore: VWR Chemicals
Descrizione: High quality Cone and Plate viscosity standards are ideal for the paints and coatings industry.
Fornitore: VWR Chemicals
Descrizione: High Temperature viscosity standards are ideal for the calibration and verification of glass capillary viscometers and other viscosity measuring equip...
Fornitore: Thermo Scientific
Descrizione: Viscometry Standards, Viscosità a 40 °C: 930±6 cSt, Viscosità a 50 °C: 82 cSt
Fornitore: PerkinElmer
Descrizione: Viscosity standard reference kit, 5×500 ml, Viscometry Standards, Hydrocarbons
Prezzo a richiesta
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Questo prodotto è stato bloccato dalla vostra organizzazione. Si prega di contattare il vostro responsabile acquisti per ulteriori informazioni.
Il prodotto originale non è più disponibile. L'articolo suggerito è disponibile
Il prodotto marcato con questo simbolo non è più disponibile o vendibile fino ad esaurimento scorte. Alternative possono essere disponibili utilizzando il numero di catalogo VWR. Per maggiori informazioni può contattare il nostro customer service al 023320311 o inviando una email all'indirizzo ebusiness.it@vwr.com
1 - 16 of 20
no targeter for Bottom