Stampa…
Accessories, WHEATON®
Fornitore: DWK Life Sciences
720-0709EA 88.8 EUR
720-0709
Accessories, WHEATON®
Contenitori Vaschette di colorazione
PP, orange.
Polypropylene grip allows for easy and safe transfer of five slides to other containers for staining. It fits into Wheaton Coplin staining jars.