/
Italy
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
VWR®, Cleanroom Disposable Goggles
111-0615
:
VWR Collection
VWR®, Cleanroom Disposable Goggles
111-0615
:
VWR Collection
68,10 €
1 * 10 pezzi
-
+
Type:
Cleanroom, sterile, anti-fog, anti-scratch
Colour:
Transparent
Lenses:
Clear PC
Pk:
10