Stampa…
SafetyCaps for Merck® bottles
Fornitore: SCAT Europe
554-3493EA 122 EUR
554-3493
SafetyCaps for Merck® bottles
Ghiere Tappi di sicurezza e per rifiuti
Use SafetyCaps without an adapter, directly on supply bottles with thread sizes S 40 and GL 40. Many solvents are already supplied in such containers. With these SafetyCaps, you can connect the containers directly to the HPLC system, without decanting or thread adapters.