1-Butyl-1-methylpyrrolidiniumtrifluormethansulphonate, Sigma-Aldrich®
Fornitore: Merck
M-Clarity™ quality level = MQ200
4.91029.0500EA
6430
EUR
4.91029.0500
4.91029.0100
1-Butyl-1-methylpyrrolidiniumtrifluormethansulphonate, Sigma-Aldrich®
1-Butyl-1-methylpyrrolidiniumtrifluormethansulphonate
Formula:
C₁₀H₂₀F₃NO₃S Peso molecolare: 291,33 g/mol |
MDL Number:
MFCD08458925 CAS Number: 367522-96-1 |
Risultati del test di specificazione
Melting Point | 3 °C |
Molar Mass | 291.34 g/mol |
Density | 1.25 g/cm3 (20 °C) |
pH | 6 (H2O, 25 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...