


Butylmethylamine, Sigma-Aldrich®
Catalog # 8.18639.1500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1,5 kg
- Seq:4046
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-75 °C
- Molar Mass:87.17 g/mol
- Boiling Point:90 - 92 °C (1013 hPa)
- Flash Point:-4 °C
- Refractive Index:1.4011 (20 °C, 589 nm)
- Explosion Limit:1.3 %(V)
- Density:0.74 g/cm3 (20 °C)
Specification Test Results
About this item
:
:87.16 g/mol
ADR:3 (8),II
:M-Clarity™ quality level = MQ200
EINECS:203-791-2
UN:2945
:GHS02
:GHS06
:GHS05
Storage Temperature:Ambient
CAS Number:110-68-9
Formula:C₅H₁₃N
MDL Number:MFCD00009426