Stampa…
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide, Sigma-Aldrich®
Fornitore: Merck
M-Clarity™ quality level = MQ200
4.91046.0025EA 191 EUR
4.91046.0025 4.91046.0100
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide, Sigma-Aldrich®
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide
Formula: C₁₁H₂₀F₆N₂O₄S₂
Peso molecolare: 422,41 g/mol
MDL Number: MFCD07784447
CAS Number: 223437-11-4
Visualizza i codici
Melting Point -6 °C
Molar Mass 422.41 g/mol
Density 1.40 g/cm3
pH 5 (H2O, 20 °C)