Graduated pipettes
Supplier: Bürkle
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BURK7550-0010EA 20.3 GBP
BURK7550-0010
Graduated pipettes
Pipettes Graduated Pipettes
PP, transparent.
  • For training purposes, in industrial labs, during field research and applications where pipettes made of glass cannot be used, example in the fluorine and silicate chemistry
  • Blue, permanently coloured graduation and labelling
Order Now
Learn more