Cellulose chromatography papers
Chromatography papers are not only ideal for chromatography, but also for routine and repetitive separations, routine analysis of proteins in serum (e.g. human albumin) and several absorption applications like those common in the life sciences and diagnostics. They are made of cotton linters with more than 98% alpha-cellulose content and <0,04% ash content.
- Catalog No:
- SARTFT2-527460570N
- SARTFT-3-526-025
- 552-0374
- 552-0373
- SARTFT-2507580600K
- SARTFT1-5270190100
- SARTFT2-527580600K
- SARTFT2504-580600N
- SARTFT2503-580600N
- SARTFT2503-250250N
- SARTFT-2503300580N
- Grade:
- FN 100
- FN 30
- FN 100
- FN 100
- FN 7
- FN 100
- FN 100
- FN 4
- FN 3
- FN 3
- FN 3
- Thickness:
- 0,35 mm
- 0,9 mm
- 0,35 mm
- 0,35 mm
- 0,32 mm
- 0,35 mm
- 0,35 mm
- 0,24 mm
- 0,19 mm
- 0,19 mm
- 0,19 mm
- Weight:
- 195 g/m²
- 320 g/m²
- 195 g/m²
- 195 g/m²
- 150 g/m²
- 195 g/m²
- 195 g/m²
- 120 g/m²
- 90 g/m²
- 90 g/m²
- 90 g/m²
- Capillary rise:
- 115 mm/30 min
- 240 mm/30 min
- 115 mm/30 min
- 115 mm/30 min
- 145 mm/30 min
- 115 mm/30 min
- 115 mm/30 min
- 95 mm/30 min
- 95 mm/30 min
- 95 mm/30 min
- 95 mm/30 min
Grade FN 3
- SizePk
- SheetsSheetsSARTFT2503-250250NFT2503-250250N250×250 mm1001 * 100 pezzi208,00 €
Specifications
GradeFN 3Environmentally PreferableThickness0,19 mmWeight90 g/m²Capillary rise95 mm/30 min - SheetsSheetsSARTFT-2503300580NFT-2503300580N300×580 mm1001 * 100 pezzi478,00 €
Specifications
GradeFN 3Environmentally PreferableThickness0,19 mmWeight90 g/m²Capillary rise95 mm/30 min - SheetsSheetsSARTFT2503-580600NFT2503-580600N580×600 mm1001 * 100 pezzi308,00 €
Specifications
GradeFN 3Environmentally PreferableThickness0,19 mmWeight90 g/m²Capillary rise95 mm/30 min
Grade FN 4
- SizePk
- SheetsSheetsSARTFT2504-580600NFT2504-580600N580×600 mm1001 * 100 pezzi605,00 €
Specifications
GradeFN 4Environmentally PreferableThickness0,24 mmWeight120 g/m²Capillary rise95 mm/30 min
Grade FN 7
- SizePk
- SheetsSheetsSARTFT-2507580600KFT-2507580600K580×600 mm501 * 50 pezzi335,00 €
Specifications
GradeFN 7Environmentally PreferableThickness0,32 mmWeight150 g/m²Capillary rise145 mm/30 min
Grade FN 30
- SizePk
- DiscsDiscsSARTFT-3-526-025FT-3-526-02525 mm1001 * 100 pezzi29,00 €
Specifications
GradeFN 30Environmentally PreferableThickness0,9 mmWeight320 g/m²Capillary rise240 mm/30 min
Grade FN 100
- SizePk
- SheetsSheetsSARTFT2-527460570NFT2-527460570N460×570 mm1001 * 100 pezzi648,00 €
Specifications
GradeFN 100Environmentally PreferableThickness0,35 mmWeight195 g/m²Capillary rise115 mm/30 min - RollRollSARTFT1-5270190100FT1-5270190100190 mm ×100 m11 * 1 pezzi454,00 €
Specifications
GradeFN 100Environmentally PreferableThickness0,35 mmWeight195 g/m²Capillary rise115 mm/30 min - SheetsSheets552-0373FT-2527200200N200×200 mm1001 * 100 pezzi149,00 €
Specifications
GradeFN 100Environmentally PreferableThickness0,35 mmWeight195 g/m²Capillary rise115 mm/30 min - SheetsSheets552-0374FT2-527460570K460×570 mm501 * 50 pezzi328,00 €
Specifications
GradeFN 100Environmentally PreferableThickness0,35 mmWeight195 g/m²Capillary rise115 mm/30 min - SheetsSheetsSARTFT2-527580600KFT2-527580600K580×600 mm501 * 50 pezzi431,00 €
Specifications
GradeFN 100Environmentally PreferableThickness0,35 mmWeight195 g/m²Capillary rise115 mm/30 min