Italy
Order Entry
Calibration weights, OIML class F1, Proof Line
Calibration weights, OIML class F1, Proof Line
Supplier:  Sartorius Balances
Certificates
undefined
Calibration weights, OIML class F1, Proof Line
Supplier:  Sartorius Balances
F1 weights in plastic cases with DKD certificate
Product Information
  • Catalog #
    Supplier #
    Weight
    Tolerance
    Type
    Pk
    Availability
    Price
  • SARBYCW713-AC-02
    YCW713-AC-02
    10 kg
    50 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 1 230,00 €
    1 230,00 €
    1 * 1 pezzi
       1 230,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW223-02
    YCW223-02
    200 mg
    0,06 ± mg
    Leaf weight without certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 109,00 €
    109,00 €
    1 * 1 pezzi
       109,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW353-AC-02
    YCW353-AC-02
    5 g
    0,16 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 137,00 €
    137,00 €
    1 * 1 pezzi
       137,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW413-AC-02
    YCW413-AC-02
    10 g
    0,20 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 143,00 €
    143,00 €
    1 * 1 pezzi
       143,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW253-02
    YCW253-02
    500 mg
    0,08 ± mg
    Leaf weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 109,00 €
    109,00 €
    1 * 1 pezzi
       109,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW053-02
    YCW053-02
    5 mg
    0,020 ± mg
    Leaf weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 109,00 €
    109,00 €
    1 * 1 pezzi
       109,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW613-AC-02
    YCW613-AC-02
    1 kg
    5 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 351,00 €
    351,00 €
    1 * 1 pezzi
       351,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW323-02
    YCW323-02
    2 g
    0,12 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 159,00 €
    159,00 €
    1 * 1 pezzi
       159,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW423-AC-02
    YCW423-AC-02
    20 g
    0,25 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 149,00 €
    149,00 €
    1 * 1 pezzi
       149,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW423-02
    YCW423-02
    20 g
    0,25 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 173,00 €
    173,00 €
    1 * 1 pezzi
       173,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW013-02
    YCW013-02
    1 mg
    0,020 ± mg
    Leaf weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 109,00 €
    109,00 €
    1 * 1 pezzi
       109,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW623-AC-02
    YCW623-AC-02
    2 kg
    10 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 515,00 €
    515,00 €
    1 * 1 pezzi
       515,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW453-AC-02
    YCW453-AC-02
    50 g
    0,30 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 169,00 €
    169,00 €
    1 * 1 pezzi
       169,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW413-02
    YCW413-02
    10 g
    0,20 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 169,00 €
    169,00 €
    1 * 1 pezzi
       169,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW653-AC-02
    YCW653-AC-02
    5 kg
    25 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 714,00 €
    714,00 €
    1 * 1 pezzi
       714,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW023-02
    YCW023-02
    2 mg
    0,020 ± mg
    Leaf weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 109,00 €
    109,00 €
    1 * 1 pezzi
       109,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW313-02
    YCW313-02
    1 g
    0,10 ± mg
    Leaf weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 154,00 €
    154,00 €
    1 * 1 pezzi
       154,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW313-AC-02
    YCW313-AC-02
    1 g
    0,10 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 133,00 €
    133,00 €
    1 * 1 pezzi
       133,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW513-AC-02
    YCW513-AC-02
    100 g
    0,5 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 187,00 €
    187,00 €
    1 * 1 pezzi
       187,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW113-02
    YCW113-02
    10 mg
    0,025 ± mg
    Leaf weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 109,00 €
    109,00 €
    1 * 1 pezzi
       109,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW123-02
    YCW123-02
    20 mg
    0,03 ± mg
    Leaf weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 109,00 €
    109,00 €
    1 * 1 pezzi
       109,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW323-AC-02
    YCW323-AC-02
    2 g
    0,12 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 134,00 €
    134,00 €
    1 * 1 pezzi
       134,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW553-AC-02
    YCW553-AC-02
    500 g
    2,5 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 324,00 €
    324,00 €
    1 * 1 pezzi
       324,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW153-02
    YCW153-02
    50 mg
    0,04 ± mg
    Leaf weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 109,00 €
    109,00 €
    1 * 1 pezzi
       109,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW213-02
    YCW213-02
    100 mg
    0,05 ± mg
    Leaf weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 109,00 €
    109,00 €
    1 * 1 pezzi
       109,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW353-02
    YCW353-02
    5 g
    0,16 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 164,00 €
    164,00 €
    1 * 1 pezzi
       164,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW523-AC-02
    YCW523-AC-02
    200 g
    1,0 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 211,00 €
    211,00 €
    1 * 1 pezzi
       211,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
F1 weights in plastic cases without certificate
Product Information
  • Catalog #
    Supplier #
    Weight
    Tolerance
    Type
    Pk
    Availability
    Price
  • SARBYCW113-00
    YCW113-00
    10 mg
    0,025 ± mg
    Leaf weight
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 33,30 €
    33,30 €
    1 * 1 pezzi
       33,30 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW313-00
    YCW313-00
    1 g
    0,10 ± mg
    Knob weight
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 78,00 €
    78,00 €
    1 * 1 pezzi
       78,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW253-00
    YCW253-00
    500 mg
    0,08 ± mg
    Leaf weight
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 33,30 €
    33,30 €
    1 * 1 pezzi
       33,30 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW013-00
    YCW013-00
    1 mg
    0,020 ± mg
    Leaf weight
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 33,30 €
    33,30 €
    1 * 1 pezzi
       33,30 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW353-AC-00
    YCW353-AC-00
    5 g
    0,16 ± mg
    Knob weight without DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 60,40 €
    60,40 €
    1 * 1 pezzi
       60,40 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW413-AC-00
    YCW413-AC-00
    10 g
    0,20 ± mg
    Knob weight without DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 66,60 €
    66,60 €
    1 * 1 pezzi
       66,60 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW023-00
    YCW023-00
    2 mg
    0,020 ± mg
    Leaf weight
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 33,30 €
    33,30 €
    1 * 1 pezzi
       33,30 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW613-AC-00
    YCW613-AC-00
    1 kg
    5 ± mg
    Knob weight without DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 274,00 €
    274,00 €
    1 * 1 pezzi
       274,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW323-00
    YCW323-00
    2 g
    0,12 ± mg
    Knob weight
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 83,20 €
    83,20 €
    1 * 1 pezzi
       83,20 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW413-00
    YCW413-00
    10 g
    0,20 ± mg
    Knob weight
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 93,60 €
    93,60 €
    1 * 1 pezzi
       93,60 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW623-AC-00
    YCW623-AC-00
    2 kg
    10 ± mg
    Knob weight without DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 439,00 €
    439,00 €
    1 * 1 pezzi
       439,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW653-AC-00
    YCW653-AC-00
    5 kg
    25 ± mg
    Knob weight without DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 638,00 €
    638,00 €
    1 * 1 pezzi
       638,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW153-00
    YCW153-00
    50 mg
    0,04 ± mg
    Leaf weight
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 33,30 €
    33,30 €
    1 * 1 pezzi
       33,30 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW213-00
    YCW213-00
    100 mg
    0,05 ± mg
    Leaf weight
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 33,30 €
    33,30 €
    1 * 1 pezzi
       33,30 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW223-00
    YCW223-00
    200 mg
    0,06 ± mg
    Leaf weight
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 33,30 €
    33,30 €
    1 * 1 pezzi
       33,30 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW313-AC-00
    YCW313-AC-00
    1 g
    0,10 ± mg
    Knob weight without DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 56,20 €
    56,20 €
    1 * 1 pezzi
       56,20 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW353-00
    YCW353-00
    5 g
    0,16 ± mg
    Knob weight
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 88,40 €
    88,40 €
    1 * 1 pezzi
       88,40 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW423-00
    YCW423-00
    20 g
    0,25 ± mg
    Knob weight
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 97,80 €
    97,80 €
    1 * 1 pezzi
       97,80 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW323-AC-00
    YCW323-AC-00
    2 g
    0,12 ± mg
    Knob weight without DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 57,20 €
    57,20 €
    1 * 1 pezzi
       57,20 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW423-AC-00
    YCW423-AC-00
    20 g
    0,25 ± mg
    Knob weight without DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 72,80 €
    72,80 €
    1 * 1 pezzi
       72,80 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW453-AC-00
    YCW453-AC-00
    50 g
    0,30 ± mg
    Knob weight without DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 93,60 €
    93,60 €
    1 * 1 pezzi
       93,60 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW513-AC-00
    YCW513-AC-00
    100 g
    0,5 ± mg
    Knob weight without DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 110,00 €
    110,00 €
    1 * 1 pezzi
       110,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW523-AC-00
    YCW523-AC-00
    200 g
    1,0 ± mg
    Knob weight without DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 136,00 €
    136,00 €
    1 * 1 pezzi
       136,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW553-AC-00
    YCW553-AC-00
    500 g
    2,5 ± mg
    Knob weight without DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 249,00 €
    249,00 €
    1 * 1 pezzi
       249,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW053-00
    YCW053-00
    5 mg
    0,020 ± mg
    Leaf weight
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 33,30 €
    33,30 €
    1 * 1 pezzi
       33,30 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW123-00
    YCW123-00
    20 mg
    0,03 ± mg
    Leaf weight
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 33,30 €
    33,30 €
    1 * 1 pezzi
       33,30 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW713-AC-00
    YCW713-AC-00
    10 kg
    50 ± mg
    Knob weight without DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 1 150,00 €
    1 150,00 €
    1 * 1 pezzi
       1 150,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
F1 weights in wooden boxes with DKD certificate
Product Information
  • Catalog #
    Supplier #
    Weight
    Tolerance
    Type
    Pk
    Availability
    Price
  • SARBYCW613-02
    YCW613-02
    1 kg
    5 ± mg
    Knob weight with gloves and DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 490,00 €
    490,00 €
    1 * 1 pezzi
       490,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW713-02
    YCW713-02
    10 kg
    50 ± mg
    Knob weight with gloves and DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 1 570,00 €
    1 570,00 €
    1 * 1 pezzi
       1 570,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW513-02
    YCW513-02
    100 g
    0,5 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 253,00 €
    253,00 €
    1 * 1 pezzi
       253,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW453-02
    YCW453-02
    50 g
    0,30 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 203,00 €
    203,00 €
    1 * 1 pezzi
       203,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW523-02
    YCW523-02
    200 g
    1,0 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 294,00 €
    294,00 €
    1 * 1 pezzi
       294,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW553-02
    YCW553-02
    500 g
    2,5 ± mg
    Knob weight with DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 392,00 €
    392,00 €
    1 * 1 pezzi
       392,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW623-02
    YCW623-02
    2 kg
    10 ± mg
    Knob weight with gloves and DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 658,00 €
    658,00 €
    1 * 1 pezzi
       658,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW723-02
    YCW723-02
    20 kg
    100 ± mg
    Knob weight with gloves and DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 2 990,00 €
    2 990,00 €
    1 * 1 pezzi
       2 990,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW653-02
    YCW653-02
    5 kg
    25 ± mg
    Knob weight with gloves and DKD certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 1 030,00 €
    1 030,00 €
    1 * 1 pezzi
       1 030,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
F1 weights in wooden boxes without certificate
Product Information
  • Catalog #
    Supplier #
    Weight
    Tolerance
    Type
    Pk
    Availability
    Price
  • SARBYCW453-00
    YCW453-00
    50 g
    0,30 ± mg
    Knob weight without certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 126,00 €
    126,00 €
    1 * 1 pezzi
       126,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • 611-1708
    YCW623-00
    2 kg
    10 ± mg
    Knob weight with gloves, without certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 581,00 €
    581,00 €
    1 * 1 pezzi
       581,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW523-00
    YCW523-00
    200 g
    1,0 ± mg
    Knob weight without certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 217,00 €
    217,00 €
    1 * 1 pezzi
       217,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW713-00
    YCW713-00
    10 kg
    50 ± mg
    Knob weight with gloves, without certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 1 500,00 €
    1 500,00 €
    1 * 1 pezzi
       1 500,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW723-00
    YCW723-00
    20 kg
    100 ± mg
    Knob weight with gloves, without certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 2 900,00 €
    2 900,00 €
    1 * 1 pezzi
       2 900,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • 611-1707
    YCW613-00
    1 kg
    5 ± mg
    Knob weight with gloves, without certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 414,00 €
    414,00 €
    1 * 1 pezzi
       414,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW553-00
    YCW553-00
    500 g
    2,5 ± mg
    Knob weight without certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 315,00 €
    315,00 €
    1 * 1 pezzi
       315,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW513-00
    YCW513-00
    100 g
    0,5 ± mg
    Knob weight without certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 176,00 €
    176,00 €
    1 * 1 pezzi
       176,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • SARBYCW653-00
    YCW653-00
    5 kg
    25 ± mg
    Knob weight with gloves, without certificate
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 954,00 €
    954,00 €
    1 * 1 pezzi
       954,00 €
    Specifications:
    Accessories