Italy
Order Entry
3-Methylpentane
Flammable
Health
Irritant
Environment
3-Methylpentane
Catalog # PROOCHE120
CAS Number:  
Flammable
Health
Irritant
Environment
3-Methylpentane
Catalog # PROOCHE120
CAS Number:  
Price
204,00 €
Log in for availability
Unit of Measure 1 * 2 mL
Specifications
  • Pack type:
    Glass bottle
  • Pk:
    2 mL
  • Seq:
    2145
Specifications
About this item
 :
 :86.18 g/mol
Boiling Pt:64 °C (1013 hPa)
Melting Pt:–118 °C
Density:0.664 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:202-481-4
UN:1208
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:96-14-0
Formula:C₆H₁₄
MDL Number:MFCD00009342