Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Italy
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
Bromine
Bromine
Bromine
Bromine
100593P
:
VWR Chemicals
:
Bromine
100593P
:
VWR Chemicals
:
Pk:
250 mL
Seq:
2590
:
:
159.81 g/mol
:
13,01378
:
58.7 °C (1013 hPa)
:
–7.2 °C
:
3.12 g/cm³ (20 °C)
:
Ambient
:
231-778-1
:
1744
:
8,I
:
GHS06
:
GHS05
:
GHS09
:
7726-95-6
:
Br₂
:
MFCD00010896