Cone and plate viscometers
Supplier: AMETEK
Certificates
About this item
A rugged instrument range designed for measurement of medium to high viscosity samples including paints, adhesives and coatings in industrial laboratories.
- Small sample size
- Easy to use key pad and control handle for accurate automatic cone positioning
- Easy set up and cleaning
- Built-in Peltier plate for temperature control
The range comes with a choice of low (5 to 75 °C) or high (50 to 235 °C) temperature versions.
CAP1000+ with selectable standard speeds (750 or 900 min⁻¹) for QC applications and repetitive testing.
CAP2000+ with variable speed (5 to 1000 min⁻¹) for R and D as well as more detailed QC testing. Optional automated PC control with Capcalc32 software.