Stampa…
UV illuminator, FirstLight®
Fornitore: Analytik Jena
FirstLight®
732-4354EA 5910 EUR
732-4354 732-4353
UV illuminator, FirstLight®
Transilluminatori Transilluminatori a UV
The FirstLight® UV illuminator provides a highly uniform excitation source for quantitative fluorescent imaging in a wide range of genomic and proteomic applications, including electrophoretic separation and quantitation of 1D and 2D protein separations, solid phase immunoassay, and DNA and RNA quantitation. The illuminator emits 302 nm UV excitation.
  • Produces <5% coefficient of variance across the full filter area
  • Software correction of uniformity is not required
  • Uniformity ensures consistent illumination over the imaging surface, resulting in high quality images
  • Accurate gel to gel comparison is assured
W×D×H (including blocking cover): 356×279×143 mm