Italy
Order Entry
Single channel pipettes, mechanical, fixed / variable volume, Proline®
Single channel pipettes, mechanical, fixed / variable volume, Proline®
Supplier:  Sartorius
   Sustainable Options Available
Certificates
undefined
Single channel pipettes, mechanical, fixed / variable volume, Proline®
Supplier:  Sartorius
   Sustainable Options Available
Specifications
  • Catalog No:
  • 613-4054
  • 613-4087
  • 613-4077
  • 613-4088
  • 613-4078
  • 613-4079
  • 613-4045
  • 613-4053
  • 613-4080
  • 613-4047
  • 613-4081
  • 613-4046
  • 613-4050
  • 613-4082
  • 613-4052
  • 613-4083
  • 613-4084
  • 613-4051
  • 613-4085
  • 613-4086
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
Specifications
Variable volume
Product Information
  • Catalog #
    Supplier #
    Volume
    Increment
    Accuracy
    Imprecision
    Recommended tips
    With Safe-Cone filter(s)
    Availability
    Price
  • 613-4054
    720110
    1000 - 5000 µl
    50,0 µl
    ±0,70 - ±0,50 %
    <0,30 - <0,20 %
    5000 µl
    613-4068, 613-0692
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 264,00 €
    264,00 €
    1 * 1 pezzi
       264,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4045
    720015
    1 - 10 µl
    0,10 µl
    ±2,50 - ±1,00 %
    <1,50 - <0,80 %
    10, 300 / F10, F20 µl
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 264,00 €
    264,00 €
    1 * 1 pezzi
       264,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4053
    720080
    2 - 20 µl
    0,50 µl
    ±3,00 - ±0,90 %
    <2,00 - <0,40 %
    300 / F20, F100 µl
    613-4071, 613-0694
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 264,00 €
    264,00 €
    1 * 1 pezzi
       264,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4047
    720025
    5 - 50 µl
    0,50 µl
    ±2,00 - ±0,60 %
    <2,00 - <0,30 %
    300, 350 / F100 µl
    613-4071, 613-0694
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 264,00 €
    264,00 €
    1 * 1 pezzi
       264,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4046
    720010
    0,25 - 2,5 µl
    0,05 µl
    ±12,00 - ±2,50 %
    <6,00 - <2,00 %
    10 / F10 µl
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 264,00 €
    264,00 €
    1 * 1 pezzi
       264,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4050
    720050
    10 - 100 µl
    1,00 µl
    ±3,00 - ±0,80 %
    <1,00 - <0,20 %
    300, 350 / F100, F120, F200, F300 µl
    613-4070, 613-0693
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 264,00 €
    264,00 €
    1 * 1 pezzi
       264,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4052
    720070
    20 - 200 µl
    1,00 µl
    ±2,50 - ±0,60 %
    <0,80 - <0,20 %
    300, 350 / F200, F300 µl
    613-4070, 613-0693
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 264,00 €
    264,00 €
    1 * 1 pezzi
       264,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4051
    720060
    100 - 1000 µl
    5,00 µl
    ±2,00 - ±0,60 %
    <0,70 - <0,20 %
    1000 / F1000 µl
    613-4069, 613-4074
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 264,00 €
    264,00 €
    1 * 1 pezzi
       264,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
Fixed volume
Product Information
  • Catalog #
    Supplier #
    Volume
    Increment
    Accuracy
    Imprecision
    Recommended tips
    With Safe-Cone filter(s)
    Availability
    Price
  • 613-4077
    722001
    5 µl
    ±1,30 %
    <1,20 %
    10, 300 / F10, F20 µl
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 179,00 €
    179,00 €
    1 * 1 pezzi
       179,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4078
    722004
    10 µl
    ±0,80 %
    <0,80 %
    10, 300 / F10, F20 µl
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 179,00 €
    179,00 €
    1 * 1 pezzi
       179,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4079
    722010
    20 µl
    ±0,60 %
    <0,50 %
    300, 350 / F20, F100 µl
    613-4071, 613-0694
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 179,00 €
    179,00 €
    1 * 1 pezzi
       179,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4080
    722015
    25 µl
    ±0,50 %
    <0,30 %
    300, 350 / F100 µl
    613-4071, 613-0694
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 179,00 €
    179,00 €
    1 * 1 pezzi
       179,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4081
    722020
    50 µl
    ±0,50 %
    <0,30 %
    300, 350 / F100 µl
    613-4071, 613-0694
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 179,00 €
    179,00 €
    1 * 1 pezzi
       179,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4082
    722025
    100 µl
    ±0,50 %
    <0,30 %
    300, 350 / F100, F120, F200, F300 µl
    613-4070, 613-0693
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 179,00 €
    179,00 €
    1 * 1 pezzi
       179,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4083
    722030
    200 µl
    ±0,40 %
    <0,20 %
    300, 350 / F200, F300 µl
    613-4070, 613-0693
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 179,00 €
    179,00 €
    1 * 1 pezzi
       179,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4084
    722035
    250 µl
    ±0,40 %
    <0,20 %
    1000 / F500, F1000 µl
    613-4069, 613-4074
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 179,00 €
    179,00 €
    1 * 1 pezzi
       179,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4085
    722040
    500 µl
    ±0,30 %
    <0,20 %
    1000 / F500, F1000 µl
    613-4069, 613-4074
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 179,00 €
    179,00 €
    1 * 1 pezzi
       179,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4087
    722050
    2000 µl
    ±0,30 %
    <0,15 %
    5000 µl
    613-4068, 613-0692
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 179,00 €
    179,00 €
    1 * 1 pezzi
       179,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4088
    722055
    5000 µl
    ±0,30 %
    <0,15 %
    5000 µl
    613-4068, 613-0692
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 179,00 €
    179,00 €
    1 * 1 pezzi
       179,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-4086
    722045
    1000 µl
    ±0,30 %
    <0,20 %
    1000 / F1000 µl
    613-4069, 613-4074
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 179,00 €
    179,00 €
    1 * 1 pezzi
       179,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
Stands
Product Information
  • Catalog #
    Description
    For
    Colour
    W×D×H
    Pk
    Availability
    Price
  • Carousel stand for 6 mechanical pipettes
    6 mechanical pipettes
    White
    290×250×60 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 139,00 €
    139,00 €
Product Information
  • Catalog #
    Seq
    Description
    Colour
    W×D×H
    Pk
    Availability
    Price
  • 59
    Universal linear stand for holding up to 11 pipettes in an upright position
    White/black
    290×250×60 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 84,00 €
    84,00 €
Product Information
  • Catalog #
    Seq
    Description
    Colour
    Pk
    Availability
    Price
  • 60
    Holder for 1 pipette (mLINE® or Proline® Plus), can be attached to an edge of a table or a shelf
    Black