Stampa…
Temperature and humidity chart recorders
Fornitore: Thies Clima
628-2150EA 1800 EUR
628-2150 628-2152 628-2153
Temperature and humidity chart recorders
Registratori grafici Registratori
These instruments permit the recording of temperature and humidity data. The axes are supported by pivot. Mechanical drum clockwork mechanism with hand winding for the temperature range between −35 and 80 °C or quartz clockwork, battery-operated 1,5 V Mignon cell (LR6A) with switchable thrusts in the temperature range between −10 and 60 °C.
  • Recording time: 1 or 7 days and 31 days for 628-2153