Stampa…
Zero air generators
Fornitore: Parker Balston
559-0017EA 0 EUR
559-0017 PARKHPZA-3500UK PARKHPZA-30000UK 559-0014 PARK75-83UK
Zero air generators
Generatori di gas Generatori di aria zero
Designed as a compact, hazard free, lower cost alternative to the user of expensive high pressure gas cylinders in the laboratory. Replacing cylinders with a gas generator brings advantages: increased safety, greater convenience, reduced cost and higher purity gas for more accurate analysis.
  • Produces zero air from compressed air
  • Ensures a flat and consistent base line
  • Short payback period
  • Compact and reliable
Purity: <0,05 ppm THC.
Inlet pressure 2,8 to 8,6 bar.
Inlet/outlet port size: ¼" NPT female.