Stampa…
Hai cercato: Filtri gas
62  risultati sono stati trovati
SearchResultCount:"62"
Sort Results
Visualizzazione a lista Nuova Visualizzazione
La tua valutazione
Codice catalogo: (23988.)
Fornitore: Merck
Codice articolo fornitore: 23988
Codice articolo locale: SUPL23988
Descrizione: This economy water vapor trap is designed for moisture removal for compressed air service to pneumatically-controlled devices.
UOM: 1 * 1 Cad.
Fornitore: SGE Analytical Science
Descrizione: Big Trap gas purifiers are designed for bulk purification applications, or where several instruments are plumbed from a single source.
Fornitore: ERLAB (INCL. CAPTAIRE)
Descrizione: Carbon filter for 834/1634, Per: Use with formaldehyde vapours
Fornitore: ENTEGRIS INC
Descrizione: GateKeeper® GPU gas purifiers were designed to improve purity consistency, lifetime and final purity. Leveraging its expertise in material science, En...
Fornitore: ENTEGRIS INC
Descrizione: These inline gas filters are designed to removed particles from inert gases for flow rates up to 30 L/min.
Fornitore: ENTEGRIS INC
Descrizione: Wafergard® SG FG series gas filters provide superior particulate filtration to maximise photovoltaic cell and display manufacturing production yields.
Fornitore: ENTEGRIS INC
Descrizione: These filters are compatible with high purity gases as well as general nitrogen and CDA applications. Available in Bronze and Silver performance level...
Promo
Fornitore: ENTEGRIS INC
Descrizione: The patented nickel filter membrane of these inline gas filters offers superior corrosion resistance and excellent compatibility with inert and reacti...
Fornitore: SGE Analytical Science
Descrizione: Gas purifiers are designed to provide fast stabilisation times to reduce gas consumption and provide clean gas to GC and GC/MS systems.
Fornitore: SGE Analytical Science
Descrizione: A high-capacity gas purifying system that can remove a combination of oxygen, moisture and hydrocarbons, (greater than C4) from inert gases and hydrog...
Fornitore: SGE Analytical Science
Descrizione: The ZPure™ line of filters removing a wide range of contaminants to trace levels.
Fornitore: SGE Analytical Science
Descrizione: Pure gas is a crictical requirement in gas chromatography, spectroscopy, optics, lithography and numerous other applications in manufacturing and labo...
Fornitore: SGE Analytical Science
Descrizione: The ZPure™ H₂O/HC filter removes moisture and hydrocarbons from dry air or inert gases. The co-free moisture indicator signals breakthrough when it ch...
Fornitore: SGE Analytical Science
Descrizione: The ZPure™ H₂O 'High Sensitivity Moisture Indicating' Filter is used for applications requiring high efficiency moisture removal. The high sensitivity...
Fornitore: SGE Analytical Science
Descrizione: The ZPure™ H₂O filter removes moisture to low ppb levels. The Co-free moisture indicator signals breakthrough when it changes from green to blue.
Fornitore: SGE Analytical Science
Descrizione: The ZPure™ line of filters remove a wide range of contaminants to trace levels.
Prezzo a richiesta
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Questo prodotto è stato bloccato dalla vostra organizzazione. Si prega di contattare il vostro responsabile acquisti per ulteriori informazioni.
Il prodotto originale non è più disponibile. L'articolo suggerito è disponibile
Il prodotto marcato con questo simbolo non è più disponibile o vendibile fino ad esaurimento scorte. Alternative possono essere disponibili utilizzando il numero di catalogo VWR. Per maggiori informazioni può contattare il nostro customer service al 023320311 o inviando una email all'indirizzo ebusiness.it@vwr.com
1 - 16 of 62
no targeter for Bottom