Liquid nitrogen transport vessels, Voyageur
- Catalog No:
- 478-3312
- 478-3311
- 478-3314
- 478-3313
- Model:
- Voyageur 5
- Voyageur 2
- Voyageur plus
- Voyageur 12
- Capacity:
- 6,5 L
- 1,75 L
- 20,6 L
- 15 L
- Absorbed capacity:
- 4,8 L
- 1,35 L
- 7,2 L
- 10,5 L
- Static holding time (days):
- 37
- 13
- 9,5
- 44
- Daily evaporation:
- 0,13 L
- 0,1 L
- 0,8 L
- 0,24 L
- Ø neck:
- 50 mm
- 30 mm
- 215 mm
- 80 mm
- Ø×H:
- 248×550 mm
- 174×395 mm
- 356×575 mm
- 308×570 mm
- No. of canisters:
- 2
- 2
- -
- 2
- Ø×H canisters:
- 41×280 mm
- 26×120 mm
- -
- 71×280 mm
- No. of stages of goblets:
- 2
- 1
- -
- 2
- Storage capacity of straws - 0,25 ml straws (max):
- 1040
- 220
- -
- 3280
- Storage capacity of straws - 0,5 ml straws (max):
- 400
- 100
- -
- 1320
- Storage capacity of 2 ml vials - capacity (100 vials/box):
- 84
- -
- 500
- 252
- Storage capacity of 5 ml vials - capacity (100 vials/box):
- 42
- -
- 162
- 126
- Storage capacity of bags - max. DF700 bags:
- -
- -
- 6
- -
- Pk
- Voyageur 2: VOYAGEUR21: AIRLVOYAGEUR21
Accessories
Specifications
ModelVoyageur 2Capacity1,75 LEnvironmentally PreferableAbsorbed capacity1,35 LStatic holding time (days)13Daily evaporation0,1 LØ neck30 mmØ×H174×395 mmNo. of canisters2Ø×H canisters26×120 mmNo. of stages of goblets1Storage capacity of straws - 0,25 ml straws (max)220Storage capacity of straws - 0,5 ml straws (max)100Storage capacity of 2 ml vials - capacity (100 vials/box)-Storage capacity of 5 ml vials - capacity (100 vials/box)-Storage capacity of bags - max. DF700 bags- - Voyageur 5: VOYAGEUR52: AIRLVOYAGEUR52
Accessories
Specifications
ModelVoyageur 5Capacity6,5 LEnvironmentally PreferableAbsorbed capacity4,8 LStatic holding time (days)37Daily evaporation0,13 LØ neck50 mmØ×H248×550 mmNo. of canisters2Ø×H canisters41×280 mmNo. of stages of goblets2Storage capacity of straws - 0,25 ml straws (max)1040Storage capacity of straws - 0,5 ml straws (max)400Storage capacity of 2 ml vials - capacity (100 vials/box)84Storage capacity of 5 ml vials - capacity (100 vials/box)42Storage capacity of bags - max. DF700 bags- - Voyageur 12Voyageur 12478-3313VOYAGEUR12211 * 1 pezzi2 110,00 €: VOYAGEUR122: AIRLVOYAGEUR122
Accessories
Specifications
ModelVoyageur 12Capacity15 LEnvironmentally PreferableAbsorbed capacity10,5 LStatic holding time (days)44Daily evaporation0,24 LØ neck80 mmØ×H308×570 mmNo. of canisters2Ø×H canisters71×280 mmNo. of stages of goblets2Storage capacity of straws - 0,25 ml straws (max)3280Storage capacity of straws - 0,5 ml straws (max)1320Storage capacity of 2 ml vials - capacity (100 vials/box)252Storage capacity of 5 ml vials - capacity (100 vials/box)126Storage capacity of bags - max. DF700 bags- - Voyageur plusVoyageur plus478-3314VOYAGEUR20211 * 1 pezzi3 320,00 €: VOYAGEUR202: AIRLVOYAGEUR202
Accessories
Specifications
ModelVoyageur plusCapacity20,6 LEnvironmentally PreferableAbsorbed capacity7,2 LStatic holding time (days)9,5Daily evaporation0,8 LØ neck215 mmØ×H356×575 mmNo. of canisters-Ø×H canisters-No. of stages of goblets-Storage capacity of straws - 0,25 ml straws (max)-Storage capacity of straws - 0,5 ml straws (max)-Storage capacity of 2 ml vials - capacity (100 vials/box)500Storage capacity of 5 ml vials - capacity (100 vials/box)162Storage capacity of bags - max. DF700 bags6