Italy
Order Entry
Rotors for 5424 centrifuge
 :  EPPENDORF
undefined
Rotors for 5424 centrifuge
 :  EPPENDORF
  • Catalog No:
  • 521-0741
  • 521-0737
  • 521-0739
  • 521-0742
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  •  
    Pk
  • Fixed angle rotor FA-45-24-11-Special, 24×1,5/2 ml, PTFE coated, 20238 ×g, incl. lid
    Fixed angle rotor FA-45-24-11-Special, 24×1,5/2 ml, PTFE coated, 20238 ×g, incl. lid
    521-0737
    5424700.004
    1
    1 * 1 pezzi
    1 * 1 pezzi - 932,00 €
    932,00 €
    1 * 1 pezzi
       932,00 €
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Fixed angle rotor FA-45-24-11, 24×1,5/2 ml, 20238 ×g, incl. lid
    Fixed angle rotor FA-45-24-11, 24×1,5/2 ml, 20238 ×g, incl. lid
    521-0739
    5424702.007
    1
    1 * 1 pezzi
    1 * 1 pezzi - 738,00 €
    738,00 €
    1 * 1 pezzi
       738,00 €
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Fixed angle rotor F-45-18-11-Kit with extended rim, 18×1,5/2 ml, 17347 ×g, incl. lid
    Fixed angle rotor F-45-18-11-Kit with extended rim, 18×1,5/2 ml, 17347 ×g, incl. lid
    521-0742
    5424706.002
    1
    1 * 1 pezzi
    1 * 1 pezzi - 583,00 €
    583,00 €
    1 * 1 pezzi
       583,00 €
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Fixed angle rotor F-45-32-5-PCR, 4×8 PCR tubes, 17829 ×g, incl. lid
    Fixed angle rotor F-45-32-5-PCR, 4×8 PCR tubes, 17829 ×g, incl. lid
    521-0741
    5424704.000
    1
    1 * 1 pezzi
    1 * 1 pezzi - 526,00 €
    526,00 €
    1 * 1 pezzi
       526,00 €
      Low Manufacturing Impact
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact