Tubing selector
Catalog # 229-0111
Supplier: KARTELL
- Description:Tubing selector
- L×W:165×90 mm
- Pk:1
ABS, green.
- Sturdy, handy shape
- Engraved numbers for easy reading
For rapid determination of the internal and external diameters of tubing, glass tubes, glass rods, test tubes etc. For internal diameters from 1 to 25 mm and external diameters from 1,5 to 21 mm.