Italy
Order Entry
Microcentrifuge tubes, 2,0 ml
  211-2606
 :  Brand
undefined
Microcentrifuge tubes, 2,0 ml
  211-2606
 :  Brand
41,90 €
  1 * 500 pezzi
  • Type:
    with attached lid
  • Version:
    graduated, conical bottom, non sterile
  • Capacity:
    2,0 ml
  • Colour:
    Colourless
  • Pk:
    500
PP, graduated, autoclavable with attached cap.
  • Can withstand centrifugation up to 20000×g
  • Flat cap can be easily labelled and pierced
  • Frosted marking area