Stampa…
Hai cercato: Carrelli
98  risultati sono stati trovati
SearchResultCount:"98"
Sort Results
Visualizzazione a lista Nuova Visualizzazione
La tua valutazione
Fornitore: VWR Collection
Descrizione: Carrelli, Trolley, 600×400, 150 kg load capacity, Capacità di carico: 150 kg, L×P×A: 620×420×170 mm
Fornitore: VWR Collection
Descrizione: Standard stainless steel trolleys are designed to support Avantor® support containers.
Fornitore: Metro International
Descrizione: MetroMax i® glassware carts are available in two and four basket capacities and are ideal for transporting glassware safely in the laboratory.
Fornitore: MULTIROIR
Descrizione: These multi-use trolleys have been designed for intensive use. They are made of an aluminium frame with heavy duty polypropylene trays.
Codice catalogo: (141-1389)
Fornitore: PFENNIG REINIGUNGSTECHNIK
Codice articolo fornitore: 2099970
Codice articolo locale: PPSA2099970
Descrizione: The Clino® Profi C9 ergonomically optimised system trolley is exclusively developed and used for EasyMop-technique. It is made of stainless steel.
UOM: 1 * 1 pezzi
Codice catalogo: (141-0510)
Fornitore: MIELE
Codice articolo fornitore: 6392900
Codice articolo locale: MIEL6392900
Descrizione: Carrello da trasporto per il carico e lo scarico dei carrelli per lavaggio.
UOM: 1 * 1 pezzi
Fornitore: Metro International
Descrizione: These steel or chrome plated carts consist of Super Erecta shelves with plastic split sleeves, two one-piece handles of matching finish, and designate...
Fornitore: Metro International
Descrizione: myCart™ series utility carts are made from corrosion proof polymer shelves and are suitable for daily use.
Fornitore: PFENNIG REINIGUNGSTECHNIK
Descrizione: The STEC ergonomically optimised system trolley is exclusively developed and used for controlled areas. It is made of stainless steel, has enclosed ap...
Fornitore: PFENNIG REINIGUNGSTECHNIK
Descrizione: Carrelli, Dual-bucket trolley, Clino® CR5 FP-GMP, in stainless steel, with Ringo CR flat wringer, 3 buckets and waste material unit 2×70 l - for areas with the highest requirements on GMP conformity, Colore: grigio, Materiale: Acciaio inossidabile, L×P×A: 1280×500×1030 mm
Promo
Fornitore: asecos
Descrizione: Tipping skips are equipped with subframe with inserts for transportation by forklift truck. Releasing the locking device immediately activates the tip...
Fornitore: Palbam
Descrizione: This cart is specially designed for transporting particle sensing machines around the cleanroom environment. Stainless steel 304 with electropolished ...
Fornitore: Bohlender
Descrizione: Made of aluminium profiles and polyamide connectors.
Fornitore: Metro International
Descrizione: Telaio in acciaio cromato con ripiani in HDPE a forma di vaschetta
Fornitore: DÜPERTHAL GMBH & CO
Descrizione: These pallets are used for the safe, economic and effortless transport of 1 to 8 gas cylinders with hot-dip galvanised surface.
Codice catalogo: (MSGA13267)
Fornitore: asecos
Codice articolo fornitore: 13267
Codice articolo locale: MSGA13267
Descrizione: This trolley made of PE can equip up to 4 cylinders with a load capacity of 300 kg. Safe transport is ensured with the provision for forklift.
UOM: 1 * 1 pezzi
Prezzo a richiesta
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Questo prodotto è stato bloccato dalla vostra organizzazione. Si prega di contattare il vostro responsabile acquisti per ulteriori informazioni.
Il prodotto originale non è più disponibile. L'articolo suggerito è disponibile
Il prodotto marcato con questo simbolo non è più disponibile o vendibile fino ad esaurimento scorte. Alternative possono essere disponibili utilizzando il numero di catalogo VWR. Per maggiori informazioni può contattare il nostro customer service al 023320311 o inviando una email all'indirizzo ebusiness.it@vwr.com
1 - 16 of 98
no targeter for Bottom