Italy
Order Entry
Sealing tape, PTFE
Sealing tape, PTFE
Catalog # 291-1411
Supplier:  AMMERFLON
undefined
Sealing tape, PTFE
Catalog # 291-1411
Supplier:  AMMERFLON
Price
4,70 €
Log in for availability
Unit of Measure 1 * 1 pezzi
Specifications
  • Width:
    12,0 mm
  • Description:
    Sealing tape, 12,0 m
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
This white tape is 0,1 mm thick and is used to seal threads. It is packed as a roll with protective cover for clean storage.
  • Heat-resistant from −150 to +260 °C
  • Excellent chemical resistance
  • Swell-resistant, allowing easy removal from the connection point even after continuous use