Stampa…
Hai cercato: Chiusure di sicurezza
97  risultati sono stati trovati
SearchResultCount:"97"
Sort Results
Visualizzazione a lista Nuova Visualizzazione
La tua valutazione
Fornitore: Brady
Descrizione: These robust padlocks withstand severe physical abuse and come with a hardened steel shackle making them extremely difficult to cut. They are ideal fo...
Fornitore: Brady
Descrizione: These padlocks offer the same features as the standard safety padlocks. Though specifically designed for electrical lockout applications, they are sma...
Fornitore: Brady
Descrizione: These compact and lightweight padlocks are ideal for use with all lockout equipment. The insulated key chamber protects workers from shocks when the k...
Fornitore: Brady
Descrizione: These chains are far more secure than steel cables. They are supplied with a fabric sleeve which will protect paintwork and chrome from damage.
Fornitore: Brady
Descrizione: These breaker blockers are ideal for locking out oversized and irregularly shaped switches.
Fornitore: Brady
Descrizione: These plug valve lockouts easily and effectively secure a manually-actuated plug valve.
Fornitore: Brady
Descrizione: These durable padlocks feature solid brass bodies and shackles that resist corrosion and sparks.
Codice catalogo: (BRDY045629)
Fornitore: Brady
Codice articolo fornitore: 045629
Codice articolo locale: BRDY045629
Descrizione: This lockout made of red PP prevents access to the main cylinder valve. It is manufactured to accommodate neck rings up to 88 mm in diameter.
UOM: 1 * 1 pezzi
Codice catalogo: (118-0533)
Fornitore: Brady
Codice articolo fornitore: 236916
Codice articolo locale: BRDY236916
Descrizione: This non conductive nylon hasp can accommodate up to six locks, withstands up to 450 kg, yet only weighs 30 g.
UOM: 1 * 1 pezzi
Fornitore: Brady
Descrizione: Equipping employees with the proper lockout tools and warning devices can save lives, reduce lost employee time, and cut insurance costs.
New Product
Fornitore: Brady
Descrizione: Manufactured with solid aluminium body and shackles, these padlocks are amazingly lightweight and rugged.
Fornitore: Brady
Descrizione: These durable padlocks feature solid brass bodies and stainless steel or hardened steel shackles that resist corrosion and sparks.
Fornitore: Brady
Descrizione: These miniature circuit breakers are a safe and effective method for locking out European circuit breakers as they fit most of the existing types. Rec...
Fornitore: Brady
Descrizione: These labelled group lockout hasps are constructed from tough 5052 anodised aluminium alloy.
Fornitore: Brady
Descrizione: These lockout stations (without components) are constructed of sturdy white PS with pre-drilled holes for easy wall mounting.
Fornitore: Brady
Descrizione: These SafeKey aluminium padlocks are the ultimate locks for indoor and outdoor heavy-duty safety applications.
Prezzo a richiesta
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Questo prodotto è stato bloccato dalla vostra organizzazione. Si prega di contattare il vostro responsabile acquisti per ulteriori informazioni.
Il prodotto originale non è più disponibile. L'articolo suggerito è disponibile
Il prodotto marcato con questo simbolo non è più disponibile o vendibile fino ad esaurimento scorte. Alternative possono essere disponibili utilizzando il numero di catalogo VWR. Per maggiori informazioni può contattare il nostro customer service al 023320311 o inviando una email all'indirizzo ebusiness.it@vwr.com
1 - 16 of 97
no targeter for Bottom