Stampa…
Hai cercato: Medicazioni per Ferite
33  risultati sono stati trovati
SearchResultCount:"33"
Sort Results
Visualizzazione a lista Nuova Visualizzazione
La tua valutazione
Fornitore: B. Braun
Descrizione: Swabs off the roll, made of highly bleached cellulose dressing. For skin cleansing before injections, as an absorbent pad.
Fornitore: Plum Safety
Descrizione: Dispenser with elastic finger plasters made from breathable material. The plasters can be wrapped around the finger multiple times. The dispenser cont...
Codice catalogo: (118-0230)
Fornitore: Plum Safety
Codice articolo fornitore: 5504
Codice articolo locale: PLUM5504
Descrizione: The small and practical QuickFix Mini packs can be placed in glove compartments, tool boxes, cars, bags, pockets, first aid kits, etc.
UOM: 1 * 1 pezzi
Fornitore: Plum Safety
Descrizione: This plaster dispenser allows quick and hygienic first-aid access, at the centre of the workplace. Pull to retrieve a ready-for-use plaster, and get b...
Codice catalogo: (BRAU9080120N)
Fornitore: B. Braun
Codice articolo fornitore: 9080120N
Codice articolo locale: BRAU9080120N
Descrizione: Silk plasters for the fixation of all types of wound dressing, e.g. Askina® Pad, Askina® gauze compresses. Also suitable for the attachment of cathete...
UOM: 1 * 12 pezzi
Fornitore: B. Braun
Descrizione: Gauze compresses with cut edges for all types of classic wound care, as an ointment or medication carrier.
Fornitore: Plum Safety
Descrizione: Handy dispenser containing 60 detectable finger plasters which can be wrapped around the finger multiple times. The highly visible blue plasters with ...
Fornitore: Plum Safety
Descrizione: This small and modern plaster dispenser is available empty or with 45 elastic plasters for the treatment of minor cuts. Ideal for use in offices, rest...
Codice catalogo: (118-0549)
Fornitore: Plum Safety
Codice articolo fornitore: 5152
Codice articolo locale: PLUM5152
Descrizione: Wound dressing kit for effective first aid of both minor and heavy bleeding. For heavy bleeding, such as the rupture of a main artery, the small wound...
UOM: 1 * 3 pezzi
Fornitore: Plum Safety
Descrizione: This plaster dispenser allows quick and hygienic first-aid access, at the centre of the workplace. Pull to retrieve a ready-for-use plaster, and get b...
Fornitore: B. Braun
Descrizione: Elastic fixation bandage made from 54% polyamide and 46% viscose for securing wound dressings and splints and for light compression bandages.
Codice catalogo: (BRAU9048120)
Fornitore: B. Braun
Codice articolo fornitore: 9048120
Codice articolo locale: BRAU9048120
Descrizione: Elastic, wide-mesh fixation bandage made from 70% viscose and 30% polyamide. For all types of fixation dressings and light support bandages.
UOM: 1 * 20 pezzi
Codice catalogo: (115-2515)
Fornitore: B. Braun
Codice articolo fornitore: 9050418
Codice articolo locale: BRAU9050418
Descrizione: Highly absorbent dressing made of 100% chlorine-free bleached cellulose.
UOM: 1 * 1 kg
Fornitore: Plum Safety
Descrizione: Dispenser with elastic finger plasters made from breathable material. The dispenser contains 90 plasters (2 refills) and can be wall-mounted.
Fornitore: Plum Safety
Descrizione: This plaster dispenser allows quick and hygienic first-aid access, at the centre of the workplace. Pull to retrieve a ready-for-use plaster, and get b...
Fornitore: Plum Safety
Descrizione: Dispenser containing highly visible blue plasters with an integrated metal surface that can be detected via a metal detector. The dispenser contains 9...
Prezzo a richiesta
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Questo prodotto è stato bloccato dalla vostra organizzazione. Si prega di contattare il vostro responsabile acquisti per ulteriori informazioni.
Il prodotto originale non è più disponibile. L'articolo suggerito è disponibile
Il prodotto marcato con questo simbolo non è più disponibile o vendibile fino ad esaurimento scorte. Alternative possono essere disponibili utilizzando il numero di catalogo VWR. Per maggiori informazioni può contattare il nostro customer service al 023320311 o inviando una email all'indirizzo ebusiness.it@vwr.com
1 - 16 of 33
no targeter for Bottom