Italy
Order Entry
Syringe filters, Minisart® NML
 :  Sartorius
undefined
Syringe filters, Minisart® NML
 :  Sartorius
Minisart NML SFCA
  •  
    Ø
    Pore size
    Membrane
    Housing material
    Colour
    Inlet/outlet connections
  • 611-0691
    16534-GUK
    28 mm
    0,20 µm
    Surfactant-free Cellulose Acetate (SFCA)
    MBS
    Blue
    Female Luer Lock/Male Luer Lock
    1 * 50 pezzi
    1 * 50 pezzi - 236,00 €
    236,00 €
    1 * 50 pezzi
       236,00 €
    Specifications
    Environmentally Preferable
  • 611-0707
    17597-K
    28 mm
    0,20 µm
    Surfactant-free Cellulose Acetate (SFCA)
    MBS
    Blue
    Female Luer Lock/Male Luer Slip
    1 * 50 pezzi
    1 * 50 pezzi - 237,00 €
    237,00 €
    1 * 50 pezzi
       237,00 €
    Specifications
    Environmentally Preferable
  • 611-0708
    17597-Q
    28 mm
    0,20 µm
    Surfactant-free Cellulose Acetate (SFCA)
    MBS
    Blue
    Female Luer Lock/Male Luer Slip
    1 * 500 pezzi
    1 * 500 pezzi - 1 410,00 €
    1 410,00 €
    1 * 500 pezzi
       1 410,00 €
    Specifications
    Environmentally Preferable
  • 514-7011
    16534-K
    28 mm
    0,20 µm
    Surfactant-free Cellulose Acetate (SFCA)
    MBS
    Blue
    Female Luer Lock/Male Luer Lock
    1 * 50 pezzi
    1 * 50 pezzi - 236,00 €
    236,00 €
    1 * 50 pezzi
       236,00 €
    Specifications
    Environmentally Preferable
  • 514-7012
    16534-Q
    28 mm
    0,20 µm
    Surfactant-free Cellulose Acetate (SFCA)
    MBS
    Blue
    Female Luer Lock/Male Luer Lock
    1 * 500 pezzi
    1 * 500 pezzi - 1 410,00 €
    1 410,00 €
    1 * 500 pezzi
       1 410,00 €
    Specifications
    Environmentally Preferable
  • 514-0672
    16555-GUK
    28 mm
    0,45 µm
    Surfactant-free Cellulose Acetate (SFCA)
    MBS
    Yellow
    Female Luer lock/Male LuerLock
    1 * 50 pezzi
    1 * 50 pezzi - 236,00 €
    236,00 €
    1 * 50 pezzi
       236,00 €
    Specifications
    Environmentally Preferable
  • 514-7013
    16555-K
    28 mm
    0,45 µm
    Surfactant-free Cellulose Acetate (SFCA)
    MBS
    Yellow
    Female Luer lock/Male Luer Lock
    1 * 50 pezzi
    1 * 50 pezzi - 236,00 €
    236,00 €
    1 * 50 pezzi
       236,00 €
    Specifications
    Environmentally Preferable
  • 514-1235
    S6555-FMGUK
    28 mm
    0,45 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Yellow
    Female Luer lock/Male Luer lock
    1 * 50 pezzi
    1 * 50 pezzi - 224,00 €
    224,00 €
    1 * 50 pezzi
       224,00 €
    Specifications
    Environmentally Preferable
  • 514-1286
    S6555-FM-Q
    28 mm
    0,45 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Yellow
    Female Luer lock/Male Luer lock
    1 * 500 pezzi
    1 * 500 pezzi - 1 340,00 €
    1 340,00 €
    1 * 500 pezzi
       1 340,00 €
    Specifications
    Environmentally Preferable
  • 514-7014
    16555-Q
    28 mm
    0,45 µm
    Surfactant-free Cellulose Acetate (SFCA)
    MBS
    Yellow
    Female Luer Lock/Male Luer Lock
    1 * 500 pezzi
    1 * 500 pezzi - 1 410,00 €
    1 410,00 €
    1 * 500 pezzi
       1 410,00 €
    Specifications
    Environmentally Preferable
  • 514-1290
    S7598-FX-Q
    28 mm
    0,45 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Yellow
    Female Luer lock/Male Luer slip
    1 * 500 pezzi
    1 * 500 pezzi - 1 340,00 €
    1 340,00 €
    1 * 500 pezzi
       1 340,00 €
    Specifications
    Environmentally Preferable
  • 514-1282
    S6534-FMGUK
    28 mm
    0,20 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Blue
    Female Luer lock/Male Luer lock
    1 * 50 pezzi
    1 * 50 pezzi - 224,00 €
    224,00 €
    1 * 50 pezzi
       224,00 €
    Specifications
    Environmentally Preferable
  • 514-1283
    S6534-FMOSK
    28 mm
    0,20 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Blue
    Female Luer lock/Male Luer lock
    1 * 50 pezzi
    1 * 50 pezzi - 224,00 €
    224,00 €
    1 * 50 pezzi
       224,00 €
    Specifications
    Environmentally Preferable
  • 514-1284
    S6534-FM-Q
    28 mm
    0,20 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Blue
    Female Luer lock/Male Luer lock
    1 * 500 pezzi
    1 * 500 pezzi - 1 340,00 €
    1 340,00 €
    1 * 500 pezzi
       1 340,00 €
    Specifications
    Environmentally Preferable
  • 514-1239
    S7597-FX-Q
    28 mm
    0,20 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Blue
    Female Luer lock/Male Luer slip
    1 * 500 pezzi
    1 * 500 pezzi - 1 340,00 €
    1 340,00 €
    1 * 500 pezzi
       1 340,00 €
    Specifications
    Environmentally Preferable
  • 514-1285
    S6555-FMOSK
    28 mm
    0,45 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Yellow
    Female Luer lock/Male Luer lock
    1 * 50 pezzi
    1 * 50 pezzi - 224,00 €
    224,00 €
    1 * 50 pezzi
       224,00 €
    Specifications
    Environmentally Preferable
  • 514-0674
    17598-K
    28 mm
    0,45 µm
    Surfactant-free Cellulose Acetate (SFCA)
    MBS
    Yellow
    Female Luer Lock/Male Luer Slip
    1 * 50 pezzi
    1 * 50 pezzi - 237,00 €
    237,00 €
    1 * 50 pezzi
       237,00 €
    Specifications
    Environmentally Preferable
  • 514-0673
    17598-Q
    28 mm
    0,45 µm
    Surfactant-free Cellulose Acetate (SFCA)
    MBS
    Yellow
    Female Luer Lock/Male Luer Slip
    1 * 500 pezzi
    1 * 500 pezzi - 1 410,00 €
    1 410,00 €
    1 * 500 pezzi
       1 410,00 €
    Specifications
    Environmentally Preferable
  • 514-1289
    S7598-FXOSK
    28 mm
    0,45 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Yellow
    Female Luer lock/Male Luer slip
    1 * 50 pezzi
    1 * 50 pezzi - 224,00 €
    224,00 €
    1 * 50 pezzi
       224,00 €
    Specifications
    Environmentally Preferable
  • 516-1160
    16569-K
    28 mm
    0,65 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Pink
    Female Luer lock/Male Luer slip
    1 * 50 pezzi
    1 * 50 pezzi - 205,00 €
    205,00 €
    1 * 50 pezzi
       205,00 €
    Specifications
    Environmentally Preferable
  • 516-0904
    16592-GUK
    28 mm
    0,80 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Green
    Female Luer lock/Male Luer lock
    1 * 50 pezzi
    1 * 50 pezzi - 205,00 €
    205,00 €
    1 * 50 pezzi
       205,00 €
    Specifications
    Environmentally Preferable
  • 512-3114
    16592-K
    28 mm
    0,80 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Green
    Female Luer lock/Male Luer lock
    1 * 50 pezzi
    1 * 50 pezzi - 205,00 €
    205,00 €
    1 * 50 pezzi
       205,00 €
    Specifications
    Environmentally Preferable
  • 514-1288
    S7597-FXOSK
    28 mm
    0,20 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Blue
    Female Luer lock/Male Luer slip
    1 * 50 pezzi
    1 * 50 pezzi - 224,00 €
    224,00 €
    1 * 50 pezzi
       224,00 €
    Specifications
    Environmentally Preferable
  • 512-3214
    16592-Q
    28 mm
    0,80 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Green
    Female Luer lock/Male Luer lock
    1 * 500 pezzi
    1 * 500 pezzi - 1 240,00 €
    1 240,00 €
    1 * 500 pezzi
       1 240,00 €
    Specifications
    Environmentally Preferable
  • 512-3116
    17593-K
    28 mm
    1,20 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Red
    Female Luer lock/Male Luer lock
    1 * 50 pezzi
    1 * 50 pezzi - 205,00 €
    205,00 €
    1 * 50 pezzi
       205,00 €
    Specifications
    Environmentally Preferable
  • 512-3216
    17593-Q
    28 mm
    1,20 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Red
    Female Luer lock/Male Luer lock
    1 * 500 pezzi
    1 * 500 pezzi - 1 240,00 €
    1 240,00 €
    1 * 500 pezzi
       1 240,00 €
    Specifications
    Environmentally Preferable
  • 512-3218
    17594-Q
    28 mm
    5,00 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Brown
    Female Luer lock/Male Luer lock
    1 * 500 pezzi
    1 * 500 pezzi - 1 320,00 €
    1 320,00 €
    1 * 500 pezzi
       1 320,00 €
    Specifications
    Environmentally Preferable
  • 514-1287
    S7594-FMOSK
    28 mm
    5,00 µm
    Surfactant-free cellulose acetate (SFCA)
    MBS
    Brown
    Female Luer lock/Male Luer lock
    1 * 50 pezzi
    1 * 50 pezzi - 224,00 €
    224,00 €
    1 * 50 pezzi
       224,00 €
    Specifications
    Environmentally Preferable
  • 512-3118
    17594-K
    28 mm
    5,00 µm
    Surfactant-free Cellulose Acetate (SFCA)
    MBS
    Brown
    Female Luer Lock/Male Luer Lock
    1 * 50 pezzi
    1 * 50 pezzi - 237,00 €
    237,00 €
    1 * 50 pezzi
       237,00 €
    Specifications
    Environmentally Preferable
Minisart NML Plus glass fiber 0.7 µm² + SFCA
  •  
    Ø
    Pore size
    Membrane
    Housing material
    Colour
    Inlet/outlet connections
  • 512-3106
    17823-K
    28 mm
    0,20 µm
    Surfactant-free cellulose acetate (SFCA), glass fiber (GF)
    MBS
    Blue
    Female Luer lock/Male Luer lock
    1 * 50 pezzi
    1 * 50 pezzi - 242,00 €
    242,00 €
    1 * 50 pezzi
       242,00 €
    Specifications
    Environmentally Preferable
  • 512-3206
    17823-Q
    28 mm
    0,20 µm
    Surfactant-free cellulose acetate (SFCA), glass fiber (GF)
    MBS
    Blue
    Female Luer lock/Male Luer lock
    1 * 500 pezzi
    1 * 500 pezzi - 1 450,00 €
    1 450,00 €
    1 * 500 pezzi
       1 450,00 €
    Specifications
    Environmentally Preferable
  • 512-3108
    17829-K
    28 mm
    0,45 µm
    Surfactant-free cellulose acetate (SFCA), glass fiber (GF)
    MBS
    Yellow
    Female Luer lock/Male Luer lock
    1 * 50 pezzi
    1 * 50 pezzi - 242,00 €
    242,00 €
    1 * 50 pezzi
       242,00 €
    Specifications
    Environmentally Preferable
  • 512-3208
    17829-Q
    28 mm
    0,45 µm
    Surfactant-free cellulose acetate (SFCA), glass fiber (GF)
    MBS
    Yellow
    Female Luer lock/Male Luer lock
    1 * 500 pezzi
    1 * 500 pezzi - 1 450,00 €
    1 450,00 €
    1 * 500 pezzi
       1 450,00 €
    Specifications
    Environmentally Preferable
Minisart NML Plus glass fiber 0.7 µm² + CA
  •  
    Ø
    Pore size
    Membrane
    Housing material
    Colour
    Inlet/outlet connections
  • 514-5002
    17825-Q
    28 mm
    1,20 µm
    Cellulose acetate (CA), glass fiber (GF)
    MBS
    Red
    Female Luer lock/Male Luer lock
    1 * 500 pezzi
    1 * 500 pezzi - 1 620,00 €
    1 620,00 €
    1 * 500 pezzi
       1 620,00 €
    Specifications
    Environmentally Preferable
Minisart NML Plus glass fiber 0.7 µm²
  •  
    Ø
    Pore size
    Membrane
    Housing material
    Colour
    Inlet/outlet connections
  • 512-3220
    17824-Q
    28 mm
    0,70 µm
    Glass fiber (GF)
    MBS
    White
    Female Luer lock/Male Luer lock
    1 * 500 pezzi
    1 * 500 pezzi - 1 080,00 €
    1 080,00 €
    1 * 500 pezzi
       1 080,00 €
    Specifications
    Environmentally Preferable
  • 512-3120
    17824-K
    28 mm
    0,70 µm
    Glass fiber (GF)
    MBS
    White
    Female Luer lock/Male Luer lock
    1 * 50 pezzi
    1 * 50 pezzi - 133,00 €
    133,00 €
    1 * 50 pezzi
       133,00 €
    Specifications
    Environmentally Preferable
  • SART17824-R
    17824-R
    28 mm
    0,70 µm
    Glass fiber (GF)
    MBS
    White
    Female Luer lock/Male Luer lock
    1 * 1.000 pezzi
    1 * 1.000 pezzi - 1 720,00 €
    1 720,00 €
    1 * 1.000 pezzi
       1 720,00 €
    Specifications
    Environmentally Preferable
Minisart High-Flow PES
  •  
    Ø
    Pore size
    Membrane
    Housing material
    Colour
    Inlet/outlet connections
  • 516-0679
    16533-GUK
    28 mm
    0,45 µm
    Polyethersulfone (PES)
    MBS
    Dark yellow
    Female Luer lock/Male Luer slip
    1 * 50 pezzi
    1 * 50 pezzi - 215,00 €
    215,00 €
    1 * 50 pezzi
       215,00 €
    Specifications
    Environmentally Preferable
  • 516-0902
    16537-Q
    28 mm
    0,45 µm
    Polyethersulfone (PES)
    MBS
    Dark yellow
    Female Luer lock/Male Luer lock
    1 * 500 pezzi
    1 * 500 pezzi - 1 280,00 €
    1 280,00 €
    1 * 500 pezzi
       1 280,00 €
    Specifications
    Environmentally Preferable
  • 514-0152
    16553-K
    28 mm
    0,10 µm
    Polyethersulfone (PES)
    MBS
    Lilac
    Female Luer lock/Male Luer lock
    1 * 50 pezzi
    1 * 50 pezzi - 215,00 €
    215,00 €
    1 * 50 pezzi
       215,00 €
    Specifications
    Environmentally Preferable
  • 512-3225
    16532-Q
    28 mm
    0,22 µm
    Polyethersulfone (PES)
    MBS
    Dark blue
    Female Luer lock/Male Luer lock
    1 * 500 pezzi
    1 * 500 pezzi - 1 280,00 €
    1 280,00 €
    1 * 500 pezzi
       1 280,00 €
    Specifications
    Environmentally Preferable
  • 516-0903
    16541-Q
    28 mm
    0,20 µm
    Polyethersulfone (PES)
    MBS
    Dark blue
    Female Luer lock/Male Luer slip
    1 * 500 pezzi
    1 * 500 pezzi - 1 280,00 €
    1 280,00 €
    1 * 500 pezzi
       1 280,00 €
    Specifications
    Environmentally Preferable
  • 516-0676
    16532-GUK
    28 mm
    0,22 µm
    Polyethersulfone (PES)
    MBS
    Dark blue
    Female Luer lock/Male Luer lock
    1 * 50 pezzi
    1 * 50 pezzi - 215,00 €
    215,00 €
    1 * 50 pezzi
       215,00 €
    Specifications
    Environmentally Preferable
  • 516-0428
    16533-K
    28 mm
    0,45 µm
    Polyethersulfone (PES)
    MBS
    Dark yellow
    Female Luer lock/Male Luer slip
    1 * 50 pezzi
    1 * 50 pezzi - 215,00 €
    215,00 €
    1 * 50 pezzi
       215,00 €
    Specifications
    Environmentally Preferable
  • 516-0901
    16533-Q
    28 mm
    0,45 µm
    Polyethersulfone (PES)
    MBS
    Dark yellow
    Female Luer lock/Male Luer slip
    1 * 500 pezzi
    1 * 500 pezzi - 1 280,00 €
    1 280,00 €
    1 * 500 pezzi
       1 280,00 €
    Specifications
    Environmentally Preferable
  • 516-0429
    16537-K
    28 mm
    0,45 µm
    Polyethersulfone (PES)
    MBS
    Dark yellow
    Female Luer lock/Male Luer lock
    1 * 50 pezzi
    1 * 50 pezzi - 215,00 €
    215,00 €
    1 * 50 pezzi
       215,00 €
    Specifications
    Environmentally Preferable
  • 514-7010
    16532-K
    28 mm
    0,22 µm
    Polyethersulfone (PES)
    MBS
    Dark blue
    Female Luer lock/Male Luer lock
    1 * 50 pezzi
    1 * 50 pezzi - 215,00 €
    215,00 €
    1 * 50 pezzi
       215,00 €
    Specifications
    Environmentally Preferable
  • 516-0680
    16541-K
    28 mm
    0,20 µm
    Polyethersulfone (PES)
    MBS
    Dark blue
    Female Luer lock/Male Luer slip
    1 * 50 pezzi
    1 * 50 pezzi - 215,00 €
    215,00 €
    1 * 50 pezzi
       215,00 €
    Specifications
    Environmentally Preferable
Minisart HY hydrophobic PTFE for venting and gas filtration
  •  
    Ø
    Pore size
    Membrane
    Housing material
    Colour
    Inlet/outlet connections
  • 514-1237
    S6596-FMOSK
    26 mm
    0,20 µm
    Polytetrafluorethylene (PTFE)
    MBS
    Yellow
    Female Luer lock/Male Luer lock
    1 * 50 pezzi
    1 * 50 pezzi - 224,00 €
    224,00 €
    1 * 50 pezzi
       224,00 €
    Specifications
    Environmentally Preferable
  • 514-1236
    S6596-FM-Q
    26 mm
    0,20 µm
    Polytetrafluorethylene (PTFE)
    MBS
    Yellow
    Female Luer lock/Male Luer lock
    1 * 500 pezzi
    1 * 500 pezzi - 1 340,00 €
    1 340,00 €
    1 * 500 pezzi
       1 340,00 €
    Specifications
    Environmentally Preferable
  • SART1659A-HYQ
    1659A-HYQ
    26 mm
    1,00 µm
    Polytetrafluorethylene (PTFE)
    MBS
    Clear
    Female Luer lock/Male Luer lock
    1 * 500 pezzi
    1 * 500 pezzi - 1 320,00 €
    1 320,00 €
    1 * 500 pezzi
       1 320,00 €
    Specifications
    Environmentally Preferable
Minisart Air hydrophobic PTFE venting
  •  
    Ø
    Pore size
    Membrane
    Housing material
    Colour
    Inlet/outlet connections
  • SART16596-HNK
    16596-HNK
    15 mm
    0,20 µm
    Polytetrafluorethylene (PTFE)
    MBS
    Yellow
    Female Luer lock/Needle (38×1.7 mm)
    1 * 50 pezzi
    1 * 50 pezzi - 277,00 €
    277,00 €
    1 * 50 pezzi
       277,00 €
    Specifications
    Environmentally Preferable
  • SART1757A-Q
    1757A-Q
    15 mm
    0,20 µm
    Polytetrafluorethylene (PTFE)
    MBS
    White
    Female Luer lock/Hose barb 4 - 6 mm stepped
    1 * 500 pezzi
    1 * 500 pezzi - 1 640,00 €
    1 640,00 €
    1 * 500 pezzi
       1 640,00 €
    Specifications
    Environmentally Preferable
  • SART1751A-Q
    1751A-Q
    15 mm
    0,20 µm
    Polytetrafluorethylene (PTFE)
    MBS
    Yellow
    Female Luer lock/Male Luer slip
    1 * 500 pezzi
    1 * 500 pezzi - 1 010,00 €
    1 010,00 €
    1 * 500 pezzi
       1 010,00 €