Stampa…
Hai cercato: Carta da filtro per analisi quantitative
26  risultati sono stati trovati
SearchResultCount:"26"
Sort Results
Visualizzazione a lista Nuova Visualizzazione
La tua valutazione
Fornitore: VWR Collection
Descrizione: 424: Filtrazione di solfuro di piombo, solfuro ferroso, solfuro di argento, carbonato alcalino, cromato di piombo, arsenico, antimonio, cadmio ...
Codice catalogo: (516-0316)
Fornitore: VWR Collection
Codice articolo locale: VWRI516-0316
Descrizione: Low nitrogen content filters for determination of nitrogen content in steel and iron samples dissolved in acid. Filtration of fine precipitates to dis...
UOM: 1 * 100 pezzi
Fornitore: Whatman products (Cytiva)
Descrizione: Whatman Grade 589/3 ashless filter paper from Cytiva is particularly useful in capturing very fine particulate samples such as insoluble molecules in...
Fornitore: Whatman products (Cytiva)
Descrizione: With less than 0,01% ash, Cytiva's Whatman Grade 589/1 filter papers can be used for the Blaine test in the cement industry and as an ash content tes...
Fornitore: Whatman products (Cytiva)
Descrizione: These filter papers made from high quality cotton linters have the finest retention of all ashless filter paper grades. Grade 42 is designed for gravi...
Fornitore: Whatman products (Cytiva)
Descrizione: Whatman Grade 589/2 filters from Cytiva are used in routine quantitative analyses, including analysis of aqueous suspension in paper, flour analysis,...
Fornitore: Whatman products (Cytiva)
Descrizione: Whatman Grade 54 hardened low ash quantitative filter paper for Büchner funnel and Hirsch funnel filtration enables fast vacuum suction filtration for...
Fornitore: Whatman products (Cytiva)
Descrizione: Cytiva's Whatman Grade 542 filters are acid hardened to allow high retention of fine precipitates in applications such as metal determinations and hyd...
Fornitore: Whatman products (Cytiva)
Descrizione: Cytiva's Whatman Grade 43 filters support inorganic analysis and other forms of instrumental analyses. The low ash content also makes Grade 43 appropr...
Fornitore: Whatman products (Cytiva)
Descrizione: Cytiva's Whatman Grade 540 is a high-purity, acid-treated cellulose lab filter paper. Mechanically strong Grade 540 is well suited for quantitative or...
Fornitore: Whatman products (Cytiva)
Descrizione: Cytiva's Whatman Grade 541 acid hardened cellulose filter paper features high wet-strength, high chemical resistance, and a large nominal particle ret...
Fornitore: Whatman products (Cytiva)
Descrizione: Cytiva's Whatman Grade 52 is a general-purpose filter paper for quantitative analysis in Büchner funnel and other vacuum filtrations. The acid-hardene...
Fornitore: Whatman products (Cytiva)
Descrizione: Cytiva's Whatman Grade 41 environmental pollution papers are among the fastest filter papers in the Whatman ashless range. This gravimetric filter is ...
Fornitore: Whatman products (Cytiva)
Descrizione: Easily fits into conical funnels.
Fornitore: Whatman products (Cytiva)
Descrizione: These highly glazed filter papers made from high quality cotton linters support vacuum filtration in Büchner funnels or Hirsch funnels and enable stro...
Fornitore: Whatman products (Cytiva)
Descrizione: Whatman Grade 40 high-purity filters from Cytiva's business are general-purpose papers for quantitative analysis. Grade 40 filters are suitable for a ...
Prezzo a richiesta
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Questo prodotto è stato bloccato dalla vostra organizzazione. Si prega di contattare il vostro responsabile acquisti per ulteriori informazioni.
Il prodotto originale non è più disponibile. L'articolo suggerito è disponibile
Il prodotto marcato con questo simbolo non è più disponibile o vendibile fino ad esaurimento scorte. Alternative possono essere disponibili utilizzando il numero di catalogo VWR. Per maggiori informazioni può contattare il nostro customer service al 023320311 o inviando una email all'indirizzo ebusiness.it@vwr.com
1 - 16 of 26
no targeter for Bottom