Stampa…
Hai cercato: Carta da filtro per analisi qualitative
122  risultati sono stati trovati
SearchResultCount:"122"
Sort Results
Visualizzazione a lista Nuova Visualizzazione
La tua valutazione
Fornitore: VWR Collection
Descrizione: Grado 402 e 302: Grado generico per attività qualitative. Adatto per la maggior parte dei solfuri. Determinazione del contenuto totale di grass...
Fornitore: Whatman products (Cytiva)
Descrizione: Whatman Grade 1 Qualitative Filter Papers are the most widely used filter paper for routine laboratory applications, qualitative analytical separation...
Fornitore: Whatman products (Cytiva)
Descrizione: A prepleated filter used for many routine analytical applications in different industries (e.g., particle separation from food extracts or filtration ...
Fornitore: Whatman products (Cytiva)
Descrizione: Compared with quadrant-folded filters, Cytiva's Whatman Grade 2V pre-folded filter paper exposes extra surface area, which enables a higher loading ca...
Fornitore: Whatman products (Cytiva)
Descrizione: A standard grade filter used in qualitative analytical techniques to determine and identify materials. For use in büchner funnels.
Promo
Codice catalogo: (515-8080)
Fornitore: Whatman products (Cytiva)
Codice articolo fornitore: 10343876
Codice articolo locale: WHAT10343876
Descrizione: Thick filter with medium to slow filtration speed, high wet strength, and good retention for smaller particles.
UOM: 1 * 100 pezzi
Fornitore: Whatman products (Cytiva)
Descrizione: A general qualitative filter with creped surface. Widely used in industrial laboratories and students laboratories for routine filtration.
Fornitore: Whatman products (Cytiva)
Descrizione: Grade 520bll ½ thick, pleated filter paper with high wet strength offering a very high flow rate.
Promo
Fornitore: Whatman products (Cytiva)
Descrizione: Standard Whatman grade filter paper for a variety of routine applications in food and beverage industries. Well-suited for determination of fat conten...
Fornitore: Whatman products (Cytiva)
Descrizione: Whatman Grade 114V smooth filter paper from Cytiva is wet-strengthened for high mechanical stability. Grade 114V fluted filters are suitable for recov...
Fornitore: Whatman products (Cytiva)
Descrizione: These extremely strong filter papers have a high wet strength due to the addition of a small quantity of chemically stable resin.
Fornitore: Whatman products (Cytiva)
Descrizione: Whatman Grade 4 Qualitative Filter Papers are a standard grade, high flow rate filter paper. They are useful in qualitative analytical separations whe...
Fornitore: Whatman products (Cytiva)
Descrizione: Ultra high loading capacity with a particle retention well suited for use with coarse or gelatinous precipitates.
Fornitore: Whatman products (Cytiva)
Descrizione: Cytiva's Whatman Grade 113V creped filter paper, for use with a conical filter funnel, offers fast filtration and high loading capacity for analysis i...
Fornitore: Whatman products (Cytiva)
Descrizione: Whatman Grade Shark Skin filter papers are a creped filter paper that is commonly used to remove impurities in a variety of applications.
Fornitore: Whatman products (Cytiva)
Descrizione: Grained, with medium fast flow rate and medium retention. A universal filter paper, used for the filtration of extracts, oils, beer, syrups etc. Also ...
Prezzo a richiesta
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Questo prodotto è stato bloccato dalla vostra organizzazione. Si prega di contattare il vostro responsabile acquisti per ulteriori informazioni.
Il prodotto originale non è più disponibile. L'articolo suggerito è disponibile
Il prodotto marcato con questo simbolo non è più disponibile o vendibile fino ad esaurimento scorte. Alternative possono essere disponibili utilizzando il numero di catalogo VWR. Per maggiori informazioni può contattare il nostro customer service al 023320311 o inviando una email all'indirizzo ebusiness.it@vwr.com
1 - 16 of 122
no targeter for Bottom