Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Italy
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
Laboratory Jacks
VWR®, Accessories for Laboratory Jacks
VWR®, Accessories for Laboratory Jacks
VWR®, Accessories for Laboratory Jacks
Catalog # 241-0227
Supplier:
VWR Collection
Expand image
VWR®, Accessories for Laboratory Jacks
Catalog # 241-0227
Supplier:
VWR Collection
Price
21,40 €
Log in
for availability
Unit of Measure
1 * 1 pezzi
-
+
Add to cart
Specifications
For:
245-1221, 245-1222, 245-1223, 245-1226, 245-1227, 245-1228
With:
M 10 thread
Description:
Retort rod, aluminium, 400 mm, Ø 12 mm, M10
Pk:
1