Stampa…
Save on sample storage essentials
Automate your storage workflow and save with Thermo Scientific sample storage starter packs. Choose a pack to suit your lab and help boost productivity.
Shop now
Save now
Learn more
Hai cercato: scatole per trasporto
28  risultati sono stati trovati
SearchResultCount:"28"
Sort Results
Visualizzazione a lista Nuova Visualizzazione
La tua valutazione
Fornitore: VWR Collection
Descrizione: Made of transparent virgin polypropylene which is food grade. Multi-use boxes are strong, transparent plastic containers that can be used to pack, sto...
Fornitore: VWR Collection
Descrizione: These perforated plastic stackable Euro containers are made from heavy-duty polypropylene, suitable for every industry and for any application. They p...
Fornitore: VWR Collection
Descrizione: These plastic stackable Euro containers are ideal for simple storage solutions as well as highly automated systems. Made from strong plastic these con...
Fornitore: VWR Collection
Descrizione: Supporto impermeabile isolante
Fornitore: VWR Collection
Descrizione: These sturdy, transparent plastic containers are made of translucent material. The translucent material allows users to check whether a container is e...
Fornitore: HEATHROW
Descrizione: PC, transparent.
Fornitore: MULTIROIR
Descrizione: Lightweight plastic pallets are made from recycled PP or HDPE and are a real alternative to wooden pallets.
Fornitore: MULTIROIR
Descrizione: These containers, with robust attached lids, are designed to stack when full and nest when empty. The recesses of the hinged lid ensure high stability...
Fornitore: Bohlender
Descrizione: Cartons for b.safe Politainer 20 L, L×P×A: 285×285×285 mm
Fornitore: MULTIROIR
Descrizione: These polypropylene pallet boxes are ideal for storage, transportation and preparation of goods and/or foodstuffs. Its material gives them exceptiona...
Fornitore: MULTIROIR
Descrizione: Medium pallet with safety edge in recycled polypropylene material resistant to shocks and temperature variations.
Codice catalogo: (HEAT120926)
Fornitore: HEATHROW
Codice articolo fornitore: 120926
Codice articolo locale: HEAT120926
Descrizione: Your reliable solution for the safe and convenient transportation of laboratory samples, instruments, and valuable items. Crafted to meet the stringen...
UOM: 1 * 1 pezzi
New Product
Codice catalogo: (MULC14713)
Fornitore: MULTIROIR
Codice articolo fornitore: 14713
Codice articolo locale: MULC14713
Descrizione: These transport boxes are ideal for storage and logistics of solutions.
UOM: 1 * 4 pezzi
Fornitore: BÜRKLE
Descrizione: PP, grey.
Fornitore: DELTA T
Descrizione: These extra strong insulated boxes can be used for delivery. Made from Neopor® the boxes consist of 2 different parts which can be fitted together to ...
Fornitore: DELTA T
Descrizione: The MonoBox are designed for transporting from one to three TempShells. The cooling element pairs fit inside the foam outer casing and are kept at the...
Prezzo a richiesta
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Questo prodotto è stato bloccato dalla vostra organizzazione. Si prega di contattare il vostro responsabile acquisti per ulteriori informazioni.
Il prodotto originale non è più disponibile. L'articolo suggerito è disponibile
Il prodotto marcato con questo simbolo non è più disponibile o vendibile fino ad esaurimento scorte. Alternative possono essere disponibili utilizzando il numero di catalogo VWR. Per maggiori informazioni può contattare il nostro customer service al 023320311 o inviando una email all'indirizzo ebusiness.it@vwr.com
1 - 16 of 28
no targeter for Bottom