Italy
Order Entry
Filter bags, BagPage®
Filter bags, BagPage®
Supplier:  INTERSCIENCE
Certificates
undefined
Filter bags, BagPage®
Supplier:  INTERSCIENCE
Certificates
Product Information
  • Catalog #
    Supplier #
    Capacity
    Packed
    W×L
    Pk
    Availability
    Price
  • Filter bag 100 with filter pososity: 280 microns, optimal blending volume 5 − 50 ml
    Filter bag 100 with filter pososity: 280 microns, optimal blending volume 5 − 50 ml
    INSC121025
    121025
    100 ml
    25/pack, 500/box
    180×95 mm
    500
    Log in for availability
    Log in for availability
    1 * 500 pezzi
    1 * 500 pezzi - 314,00 €
    314,00 €
    1 * 500 pezzi
       314,00 €
    Supplier Article Number: 121025
    Local Article Number: INSC121025
    Specifications:
    Specifications
    Environmentally Preferable
  • Filter bag +400 with filter porosity: 280 microns, optimal blending volume 5 − 300 ml
    Filter bag +400 with filter porosity: 280 microns, optimal blending volume 5 − 300 ml
    432-0008
    122025
    400 ml
    25/pack, 500/box
    300×190 mm
    500
    Log in for availability
    Log in for availability
    1 * 500 pezzi
    1 * 500 pezzi - 309,00 €
    309,00 €
    1 * 500 pezzi
       309,00 €
    Supplier Article Number: 122025
    Local Article Number: INSC122025
    Specifications:
    Specifications
    Environmentally Preferable
  • Filter bag 400 F with filter porosity: 63 microns, optimal blending volume 5 − 300 ml
    Filter bag 400 F with filter porosity: 63 microns, optimal blending volume 5 − 300 ml
    INSC122325
    122325
    400 ml
    25/pack, 500/box
    300×190 mm
    500
    Log in for availability
    Log in for availability
    1 * 500 pezzi
    1 * 500 pezzi - 293,00 €
    293,00 €
    1 * 500 pezzi
       293,00 €
    Supplier Article Number: 122325
    Local Article Number: INSC122325
    Specifications:
    Specifications
    Environmentally Preferable
  • Filter bag 400 XR with filter porosity: 280 microns, optimal blending volume 5 − 300 ml
    Filter bag 400 XR with filter porosity: 280 microns, optimal blending volume 5 − 300 ml
    INSC122425
    122425
    400 ml
    25/pack, 400/box
    300×190 mm
    400
    Log in for availability
    Log in for availability
    1 * 400 pezzi
    1 * 400 pezzi - 325,00 €
    325,00 €
    1 * 400 pezzi
       325,00 €
    Supplier Article Number: 122425
    Local Article Number: INSC122425
    Specifications:
    Specifications
    Environmentally Preferable
  • Filter bags 400 R, with filter porosity <250 microns, optimal blending volume 5 − 300 ml
    Filter bags 400 R, with filter porosity <250 microns, optimal blending volume 5 − 300 ml
    INSC161025
    161025
    400 ml
    25/pack, 500/box
    300×190 mm
    500
    Log in for availability
    Log in for availability
    1 * 500 pezzi
    1 * 500 pezzi - 309,00 €
    309,00 €
    1 * 500 pezzi
       309,00 €
    Supplier Article Number: 161025
    Local Article Number: INSC161025
    Specifications:
    Specifications
    Environmentally Preferable
  • Filter bag 3500 with filter porosity: 280 microns, optimal blending volume 200 − 3500 ml
    Filter bag 3500 with filter porosity: 280 microns, optimal blending volume 200 − 3500 ml
    INSC123010
    123010
    3500 ml
    10/pack, 100/box
    600×380 mm
    100
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 221,00 €
    221,00 €
    1 * 100 pezzi
       221,00 €
    Supplier Article Number: 123010
    Local Article Number: INSC123010
    Specifications:
    Specifications