Italy
Order Entry
Blender Bags
Blender Bags
undefined
Blender Bags
  •  
    Packed
    Thickness
    W×L
    Pk
  • Standard 80: for use with any 80 ml laboratory blenders, made from irradiated virgin food grade polythene
    Standard 80: for use with any 80 ml laboratory blenders, made from irradiated virgin food grade polythene
    432-0000
    STD80
    40 sachets of 50 bags
    65 µm
    100×150 mm
    2.000
    1 * 2.000 pezzi
    1 * 2.000 pezzi - 252,00 €
    252,00 €
    1 * 2.000 pezzi
       252,00 €
     : STD80
     : GRADSTD80
    Specifications
    Environmentally Preferable
  • Standard 400: quality bag designed to fit any 400 ml paddle blender machines, manufactured from extruded polymer tube using high-grade, food quality polythene
    Standard 400: quality bag designed to fit any 400 ml paddle blender machines, manufactured from extruded polymer tube using high-grade, food quality polythene
    432-0001
    STD400
    10 sachets of 50 bags
    65 µm
    180×300 mm
    500
    1 * 500 pezzi
    1 * 500 pezzi - 150,00 €
    150,00 €
    1 * 500 pezzi
       150,00 €
     : STD400
     : GRADSTD400
    Specifications
    Environmentally Preferable
  • Standard 3500: for use with any 3500 ml laboratory blenders
    Standard 3500: for use with any 3500 ml laboratory blenders
    432-0005
    STD3500
    5 sachets of 50 bags
    70 µm
    380×500 mm
    250
    1 * 250 pezzi
    1 * 250 pezzi - 242,00 €
    242,00 €
    1 * 250 pezzi
       242,00 €
     : STD3500
     : GRADSTD3500
    Specifications
    Environmentally Preferable
  • Lab 400: manufactured from a special polymer for high sampling volume testing laboratories
    Lab 400: manufactured from a special polymer for high sampling volume testing laboratories
    432-0499
    LAB400
    50 sachets of 50 bags
    50 µm
    180×300 mm
    2.500
    1 * 2.500 pezzi
    1 * 2.500 pezzi - 501,00 €
    501,00 €
    1 * 2.500 pezzi
       501,00 €
     : LAB400
     : GRADLAB400
    Specifications
    Environmentally Preferable
  • Separator 400 bag: designed to fit any 400 ml paddle blender machine ; made from polyethylene aand treated by gamma irradiation ; it has an inbuilt separator bag made from plastic mesh suspended inside
    Separator 400 bag: designed to fit any 400 ml paddle blender machine ; made from polyethylene aand treated by gamma irradiation ; it has an inbuilt separator bag made from plastic mesh suspended inside
    432-0003
    SEP400
    20 sachets of 10 bags
    65 µm
    180×300 mm
    200
    1 * 200 pezzi
    1 * 200 pezzi - 204,00 €
    204,00 €
    1 * 200 pezzi
       204,00 €
     : SEP400
     : GRADSEP400