Italy
Order Entry
Steriflip® Bottle Top Vacuum Filtration Systems, Nylon Net Filter
 :  Merck
undefined
Steriflip® Bottle Top Vacuum Filtration Systems, Nylon Net Filter
Supplier:  Merck
Related Information
Product Information
  • Catalog #
    Supplier #
    Membrane
    Capacity
    Filtration area
    Pore size
    Pk
    Availability
    Price
  • Membrane filter unit, Steriflip®, nylon net, Ø×H: 64×138 mm
    Membrane filter unit, Steriflip®, nylon net, Ø×H: 64×138 mm
    MILFSCNY00020
    SCNY00020
    Nylon net, hydrophilic
    50 ml
    7 cm²
    20,00 µm
    25
    Log in for availability
    1 * 25 pezzi
    1 * 25 pezzi - 247,00 €
    247,00 €
    1 * 25 pezzi
       247,00 €
    Supplier Article Number: SCNY00020
    Local Article Number: MILFSCNY00020
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Membrane filter unit, Steriflip®, nylon net, Ø×H: 64×138 mm
    Membrane filter unit, Steriflip®, nylon net, Ø×H: 64×138 mm
    MILFSCNY00040
    SCNY00040
    Nylon net, hydrophilic
    50 ml
    7 cm²
    40,00 µm
    25
    Log in for availability
    1 * 25 pezzi
    1 * 25 pezzi - 247,00 €
    247,00 €
    1 * 25 pezzi
       247,00 €
    Supplier Article Number: SCNY00040
    Local Article Number: MILFSCNY00040
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Membrane filter unit, Steriflip®, nylon net, Ø×H: 64×138 mm
    Membrane filter unit, Steriflip®, nylon net, Ø×H: 64×138 mm
    MILFSCNY00060
    SCNY00060
    Nylon net, hydrophilic
    50 ml
    7 cm²
    60,00 µm
    25
    Log in for availability
    1 * 25 pezzi
    1 * 25 pezzi - 247,00 €
    247,00 €
    1 * 25 pezzi
       247,00 €
    Supplier Article Number: SCNY00060
    Local Article Number: MILFSCNY00060
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Membrane filter unit, Steriflip®, nylon net, Ø×H: 64×138 mm
    Membrane filter unit, Steriflip®, nylon net, Ø×H: 64×138 mm
    MILFSCNY00100
    SCNY00100
    Nylon net, hydrophilic
    50 ml
    7 cm²
    100,00 µm
    25
    Log in for availability
    1 * 25 pezzi
    1 * 25 pezzi - 247,00 €
    247,00 €
    1 * 25 pezzi
       247,00 €
    Supplier Article Number: SCNY00100
    Local Article Number: MILFSCNY00100
    Specifications:
    Accessories