Italy
Order Entry
Chloroacetyl chloride ≥98%
Chloroacetyl chloride ≥98%
  22678.294
 :  
undefined
Chloroacetyl chloride ≥98%
  22678.294
 :  
 
 
   
  1 * 1 L

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Assay:
    Min. 98 %

 

 

 :
 :13,02074
 :105 °C (1013 hPa)
 :–22.5 °C
 :1.426 g/cm³ (20 °C)
 :>100 °C
 :201-171-6
 :1752
 :6.1,I
 :112.94 g/mol
 :GHS06
 :GHS05
 :GHS08
 :GHS09
 :79-04-9
 :C₂H₂Cl₂O
 :MFCD00000725